Skip to Content
Merck
All Photos(1)

Key Documents

RO82-W-1

92030502, human thyroid, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
biological source:
human thyroid
growth mode:
Adherent
karyotype:
2n = 60-82, 82 predominant
morphology:
Epithelial
products:
Thyroglobulin
receptors:
Not specified
relevant disease(s):
metastasis

Product Name

RO82-W-1, 92030502

biological source

human thyroid

growth mode

Adherent

karyotype

2n = 60-82, 82 predominant

morphology

Epithelial

products

Thyroglobulin

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human follicular thryoid carcinoma

Cell Line Description

Established from the metastases of a follicular carcinoma in a female patient. Tumorigenic in nude mice. The basal release of immuno-detectable thyroglobulin increased in response to thyroid-stimulating hormone (TSH). The cell line grows as a monolayer without the formation of follicles. Electronmicroscopy studies showed the presence of microvilli with interdigitations between neighbouring cells.

Application

A valuable model for the study of human follicular carcinoma.

Culture Medium

DMEM:Ham′s F12:MCDB 105 (2:1:1) + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:4 i.e. seeding at 2x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service