Skip to Content
Merck
All Photos(1)

Key Documents

SBAC

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

bovine adrenal gland (cortex)

packaging

tube of 5 μg 91021201-DNA-5UG
pkg of vial of cells 91021201-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Bovine adrenal cortex, normal

Cell Line Description

Derived from the fasciculata-reticularis zone of the adrenal cortex taken from three normal adult cows. FGF stimulates cell division. Cells are responsive to ACTH and other steroidogenic agents. Finite lifespan; 60-70 population doublings expected. Studies have indicated that this cell line produces both detectable BVDV antigens and infectious BVDV.

Application

Hormone studies

Culture Medium

Ham′s F12 with Fibroblast Growth Factor (FGF) (crude, 50ng/ml or pure, 500 pg/ml) + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service