Skip to Content
Merck
All Photos(1)

Key Documents

HL60 Ast.3

88120801, human blood, Lymphoblastoid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

HL60 Ast.3, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 88120801

biological source

human blood

packaging

tube of 5 μg 88120801-DNA-5UG
pkg of vial of cells 88120801-1VL

growth mode

Suspension

karyotype

Modal no. = 93, Hypertetraploid

morphology

Lymphoblastoid

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian promyelocytic leukaemia

Cell Line Description

Variant of the promyelocytic cell line HL60 isolated after treating cells with α-particle radiation and then cloned in medium containing 1.25% DMSO. Cells are capable of differentiating into neutrophils by induction with 1.75% DMSO.

Application

Differentiation studies

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 14
D13S317: 8,11
D16S539: 11
D5S818: 12
D7S820: 11,12
THO1: 7
TPOX: 8,11
vWA: 16

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS) + 1.25% Dimethyl Sulphoxide (DMSO).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. Do not allow the culture medium to become acidic or the cell line will differentiate.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service