Skip to Content
Merck
All Photos(1)

Key Documents

111A3

85062504, Application: Fusion partner in the production of heterohybridomas, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Bovine Heterohybridoma

packaging

tube of 5 μg 85062504-DNA-5UG
pkg of vial of cells 85062504-1VL

ImmunoDon

Jersey calf lymph node

Myeloma

53B3 (mouse x bovine heteromyeloma)

Reactivity

Application: Fusion partner in the production of heterohybridomas

technique(s)

cell culture | mammalian: suitable

isotype

Not specified

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse 53B3 x Bovine Jersey Calf lymph node

Cell Line Description

Non Ig secreting

Immunogen

Sheep red blood cells

Application

This line has been used as a HAT sensitive fusion partner in the production of heterohybridomas

Culture Medium

RPMI 1640 + 2mM glutamine + 1mM Sodium Pyruvate (NaP) + 10% Horse Serum (HS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Selection of the right fusion partner cell line is critical to obtain desired attributes of the hybridoma. Explore hybridoma fusion partners cell lines.

Related Content

Major technological advances have made the production of monoclonal antibodies quicker and more efficient. There are three established platforms for antibody discovery. We offer reagents for production of monoclonal antibody libraries using each of these techniques.

Major technological advances have made the production of monoclonal antibodies quicker and more efficient. There are three established platforms for antibody discovery. We offer reagents for production of monoclonal antibody libraries using each of these techniques.

Major technological advances have made the production of monoclonal antibodies quicker and more efficient. There are three established platforms for antibody discovery. We offer reagents for production of monoclonal antibody libraries using each of these techniques.

Major technological advances have made the production of monoclonal antibodies quicker and more efficient. There are three established platforms for antibody discovery. We offer reagents for production of monoclonal antibody libraries using each of these techniques.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service