Skip to Content
Merck
All Photos(1)

Key Documents

VERO Cell Line

84113001, African green monkey kidney, Fibroblast-like

Synonym(s):

Vero 81 Cells, VeroCCL81 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

VERO Cell Line, 84113001, from African green monkey kidney, A fibroblast-like kidney cell, non-WHO origin, Convention on International Trade in Endangered Species of Wild Fauna and Flora (CITES)

biological source

African green monkey kidney

growth mode

Adherent

karyotype

2n = 60, modal no. 58

morphology

Fibroblast-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

General description

Vero cells are anchorage-dependent cells and have wide applications in molecular and cell biology research. Vero cells have a pseudo-diploid karyotype and are preferred cell lines for the propagation of various viruses, including poliovirus, rotavirus influenza and rabies virus. These cells are useful hosts for vaccine production.

Cell Line Origin

Monkey African Green kidney

Cell Line Description

Established from the kidney of a normal adult African Green monkey. Susceptible to a wide range of viruses including polio, rubella, arboviruses and reoviruses.

Application

VERO cell lines have been used:
  • in the propagation of Chandipura virus (CHPV)
  • in the cytotoxicity assay to evaluate the antioxidant and anti-viral activities of Capsicum annuum methanolic extracts
  • for the propagation of T.cruzi
Virus studies: polio, rubella, arboviruses and reoviruses

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 1-3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37 °C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service