Skip to Content
Merck
All Photos(5)

Documents

C9409

Sigma-Aldrich

Creatinine Deiminase microbial

lyophilized powder, ≥25 units/mg protein

Synonym(s):

Creatinine Deaminase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

lyophilized powder

Quality Level

specific activity

≥25 units/mg protein

mol wt

~260 kDa

composition

Protein, ≥15% biuret

storage temp.

−20°C

Application

Creatinine Deiminase microbial has been used:
  • to immobilize aminosilylated glass beads based biosensor for ammonia/ammonium and creatinine detection in urine
  • in creating creatinine-sensing membrane for biophysical studies
  • to investigate the bioelectronic tongue for the simultaneous determination of urea, creatinine and alkaline ions in clinical samples

Creatinine deiminase has been used in a study to assess the application of a creatinine-sensitive biosensor for hemodialysis control. Creatinine deiminase has also been used in a study to investigate the bioelectronic tongue for the simultaneous determination of urea, creatinine and alkaline ions in clinical samples.

Biochem/physiol Actions

Creatinine deiminase catalyzes the hydrolysis of creatinine to methylhydantoine and ammonia.

Physical properties

Isoelectric point : 4.4
Michaelis constant : 3.5 x 10‾3M (Creatinine)
Structure : 6 subunits per mol of enzyme
Inhibitors : Ag+,Hg++, o-phenanthroline,monoiodoacetate
Optimum pH : 8.5 – 9.5
Optimum temperature : 65 – 75°C
pH Stability : pH 7.0 – 11.0 (30°C, 20hr)
Thermal stability : Below 65°C (pH 7.5, 1hr)

Unit Definition

One unit will hydrolyze 1.0 μmole of creatinine to N-methylhydantoin and NH3 per min at pH 7.5 at 37 °C in a coupled system with L-glutamic dehydrogenase.

Physical form

Lyophilized powder containing mannitol as stabilizer

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C M Huang et al.
Clinical chemistry, 34(1), 59-62 (1988-01-01)
We developed an enzymatic method for determination of 5-fluorocytosine in serum, using creatine iminohydrolase (EC 3.5.4.21), the Cobas-Bio analyzer, and an extant ammonia method. Analytical recovery (y) of drug added to serum (x) was good, with y = 0.97x-0.7, Sy.x
Hybrid biosensors for clinical analysis and fermentation control.
I Karube
Methods in enzymology, 137, 131-138 (1988-01-01)
Anne K Bendt et al.
Archives of microbiology, 181(6), 443-450 (2004-05-19)
In order to utilize different nitrogen sources and to survive situations of nitrogen limitation, microorganisms have developed several mechanisms to adapt their metabolism to changes in the nitrogen supply. In this communication, the use of creatinine as an alternative nitrogen
Z K He et al.
Analytical biochemistry, 283(2), 166-174 (2000-07-25)
A precise and sensitive working microflow titration procedure was developed to determine creatinine and ammonia in urine samples. This procedure is based on enzymatic conversion of creatinine, gas diffusional membrane separation of the released ammonia into an acid acceptor stream
Manuel Gutiérrez et al.
Biosensors & bioelectronics, 23(6), 795-802 (2007-10-13)
Urea and creatinine biosensors based on urease and creatinine deiminase, respectively, covalently immobilized onto ammonium selective electrodes, were included in an array together with sensors sensitive to ammonium, potassium and sodium. Generic sensors to alkaline ions were also included. All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service