Skip to Content
Merck
All Photos(4)

Documents

C8890

Sigma-Aldrich

4-Chloro-1-naphthol

horseradish peroxidase substrate, fluorogenic, ≥98% (GC), crystalline

Synonym(s):

4-chloronaphthalen-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC10H6OH
CAS Number:
Molecular Weight:
178.61
Beilstein:
509750
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

product name

4-Chloro-1-naphthol, crystalline

Assay

≥98% (GC)

Quality Level

form

crystalline

mp

118-121 °C (lit.)

solubility

acetone: 50 mg/mL, clear, colorless to very faintly brown

storage temp.

−20°C

SMILES string

Oc1ccc(Cl)c2ccccc12

InChI

1S/C10H7ClO/c11-9-5-6-10(12)8-4-2-1-3-7(8)9/h1-6,12H

InChI key

LVSPDZAGCBEQAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Chloro-1-naphthol has been used to visualize protein bands in western blotting. It has also been used in staining polyvinylidene fluoride (PVDF) membrane by Nakane method.

Biochem/physiol Actions

4-Chloro-1-naphthol serves as a substrate for horseradish peroxidase.

Substrates

Chromogenic peroxidase substrate useful in enzyme-linked detection procedures.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Limited proteolysis of fibrinogen by fibrinogenase from Echis multisquamatis venom
Chernyshenko VO
The Protein Journal, 34(2), 130-134 (2015)
Endothelial microparticles induce formation of platelet aggregates via a von Willebrand factor/ristocetin dependent pathway, rendering them resistant to dissociation
JIMENEZ JJ, et al.
Journal of Thrombosis and Haemostasis, 3(6), 1301-1308 (2005)
Ildikó Bódi et al.
Poultry science, 98(9), 3464-3470 (2018-11-28)
In the chicken bursa of Fabricius (BF), the interfollicular epithelium (IFE) consists of cylindrical- and cuboidal-shaped cells. Among the cylindrical-shaped epithelial cells, mucus-producing and caveolin-1 (Cav-1)-expressing cells can be distinguished. Occasionally, the cuboidal-shaped cells also express Cav-1, which suggests that
Ken Lemon et al.
Journal of virology, 89(5), 2849-2856 (2014-12-30)
Human respiratory syncytial virus (HRSV) is the most important viral cause of severe respiratory tract disease in infants. Two subgroups (A and B) have been identified, which cocirculate during, or alternate between, yearly epidemics and cause indistinguishable disease. Existing in
A Esen et al.
Analytical biochemistry, 132(2), 462-467 (1983-07-15)
A simple method for the immunochemical assay of zein and other prolamins is described. Alcohol-solubilized zein is spotted onto chromatography paper disks or strips, incubated with antiserum (30-60 min), washed with several changes of Tris-buffered saline (10 min), incubated with

Articles

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service