Skip to Content
Merck
All Photos(1)

Key Documents

C8737

Sigma-Aldrich

Anti-Caspr2 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-Contactin-Associated Protein 2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

mol wt

antigen 180 kDa

species reactivity

mouse, rat, human

technique(s)

immunohistochemistry: suitable
western blot: 1:200 using rat brain membranes

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

CASPR2 (Contactin-associated protein-like 2) belongs to CASPR family and neurexin superfamily.

Immunogen

peptide corresponding to amino acids residues 1315-1331 of human Caspr2.

Application

Anti-CASPR2 antibody has been used for immunohistochemitry and western blotting of proteins from mouse brain samples. It is also suitable for western blot analysis at a dilution of 1:200 by using rat brain membranes.

Biochem/physiol Actions

CASPR2 (Contactin-associated protein-like 2) associates shaker like K+ channels in the juxtaparanodal region flanking nodes of Ranvier. It is expressed in different and distinct regions of the central nervous system (CNS) where it may plays an important role in cell recognition. CASPR and CASPR2 may interact with the actin-based cytoskeleton through cytoskeleton-associated protein 4.1B. CASPR2 is crucial for synapse formation and outgrowth of axon and dendrite. CASPR2 forms a complex with G-protein-coupled receptor 37 (GPR37) and multiple PDZ domain protein 1 (MUPP1). Disruption of the complex is linked with autism spectrum disorder (ASD). Genetic variations in CASPR2 are also associated with intellectual disability, epilepsy, schizophrenia and language disorders. It also regulates diet-induced obesity.

Physical form

Lyophilized from 0.4 mg/ml in phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin, 5% sucrose, and ≤0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuko Tanabe et al.
Journal of neurochemistry, 134(4), 783-793 (2015-05-16)
Autism spectrum disorder (ASD) is a developmental brain disorder. Mutations in synaptic components including synaptic adhesion molecules have been found in ASD patients. Contactin-associated protein-like 2 (CASPR2) is one of the synaptic adhesion molecules associated with ASD. CASPR2 forms a
David A Buchner et al.
Mammalian genome : official journal of the International Mammalian Genome Society, 23(7-8), 431-442 (2012-07-04)
Despite considerable effort, the identification of genes that regulate complex multigenic traits such as obesity has proven difficult with conventional methodologies. The use of a chromosome substitution strain-based mapping strategy based on deep congenic analysis overcame many of the difficulties
Natalia Denisenko-Nehrbass et al.
The European journal of neuroscience, 17(2), 411-416 (2003-01-25)
Caspr/paranodin, a neuronal transmembrane glycoprotein, is essential for the structure and function of septate-like paranodal axoglial junctions at nodes of Ranvier. A closely related protein, Caspr2, is concentrated in juxtaparanodal regions where it associates indirectly with the shaker-type potassium channels.
Betul Bakkaloglu et al.
American journal of human genetics, 82(1), 165-173 (2008-01-09)
Autism spectrum disorders (ASD) are a group of related neurodevelopmental syndromes with complex genetic etiology. We identified a de novo chromosome 7q inversion disrupting Autism susceptibility candidate 2 (AUTS2) and Contactin Associated Protein-Like 2 (CNTNAP2) in a child with cognitive
Ivo Spiegel et al.
Molecular and cellular neurosciences, 20(2), 283-297 (2002-07-03)
The NCP family of cell-recognition molecules represents a distinct subgroup of the neurexins that includes Caspr and Caspr2, as well as Drosophila Neurexin-IV and axotactin. Here, we report the identification of Caspr3 and Caspr4, two new NCPs expressed in nervous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service