Skip to Content
Merck
All Photos(1)

Key Documents

C7971

Sigma-Aldrich

Cilostamide

phosphodiesterase inhibitor

Synonym(s):

6-[3-(N-Cyclohexyl-N-methylcarbamoyl)propoxy]quinolin-2[1H]-one, OPC 3689

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H26N2O3
CAS Number:
Molecular Weight:
342.43
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97% (HPLC)

form

powder

solubility

DMSO: 20 mg/mL, clear, colorless to faintly yellow

SMILES string

CN(C1CCCCC1)C(=O)CCCOc2ccc3NC(=O)C=Cc3c2

InChI

1S/C20H26N2O3/c1-22(16-6-3-2-4-7-16)20(24)8-5-13-25-17-10-11-18-15(14-17)9-12-19(23)21-18/h9-12,14,16H,2-8,13H2,1H3,(H,21,23)

InChI key

UIAYVIIHMORPSJ-UHFFFAOYSA-N

Biochem/physiol Actions

PDE3 catalyzes the phosphodiester bond hydrolysis, observed in secondory messengers such as cyclic adenosine monophosphate (cAMP) and cyclic guanosine monophosphate (cGMP). Inhibitors of PDE3 are generally inotropic and vasodilatory. They are being recognized for its use in treating heart failures. Cilostamide is known to delay meiotic progression.
Selective inhibitor of PDE3 (cGMP-inhibited phosphodiesterase); IC50 = 70 ± 9 nM. This inhibitory effect increases intracellular cAMP and inhibits platelet aggregation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

K M Tang et al.
European journal of pharmacology, 268(1), 105-114 (1994-06-15)
Ultraviolet irradiation of human platelet cytosol in the presence of 32P-labelled cyclic GMP (cGMP) can specifically label 110, 80, 55, 49 and 38 kDa proteins; the 110 kDa species is the subunit of cGMP-inhibited phosphodiesterase (PDE III) and the 80
L Vanhoutte et al.
Human reproduction (Oxford, England), 24(8), 1946-1959 (2009-04-28)
Controlling nuclear maturation during oocyte culture might improve nuclear-cytoplasmic maturation synchrony. In the present study, the quality of mouse and human cumulus-enclosed oocytes (CEOs) was examined after a two-step culture consisting of a three-dimensional prematuration culture (3D-PMC), followed by in
Alberto M Luciano et al.
Biology of reproduction, 85(6), 1252-1259 (2011-08-06)
Oocyte development is characterized by impressive changes in chromatin structure and function in the germinal vesicle (GV) that are crucial in conferring to the oocyte meiotic and developmental competence. During oogenesis, oocyte and follicular cells communicate by paracrine and junctional
Xiaofei Zhu et al.
BMC urology, 17(1), 54-54 (2017-07-09)
It has been shown that hosphodiesterases (PDEs) play an important role in mediating the smooth muscle tone of rat urinary bladder. However, the gene expression profiles of them were still unknown. Urinary bladder Strips were obtained from both neonatal and
Effects of cilostamide and forskolin on the meiotic resumption and embryonic development of immature human oocytes
Shu Y, et al.
Human Reproduction, 23(3), 504-513 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service