Skip to Content
Merck
All Photos(4)

Key Documents

C7423

Sigma-Aldrich

Monoclonal Anti-CD8 antibody produced in mouse

clone UCHT-4, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Monoclonal Anti-CD8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

UCHT-4, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

flow cytometry: 5 μL using 1 × 106 cells

isotype

IgG2a

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Monoclonal Anti-Human CD8 (mouse IgG2a isotype) is derived from the hybridoma produced by the fusion of mouse myeloma cell line NS-1 and splenocytes from BALB/c mice immunized with human thymocytes followed by peripheral blood T cells. CD8 (cluster of differentiation 8), is a dimeric protein, encoded by the CD8A gene mapped to human chromosome 2p12.

Specificity

Recognizes the CD8 human T cytotoxic/suppressor lymphocyte cell surface glycoprotein. Approximately 90% of thymocytes are stained in suspension. In thymus sections, both cortical and medullary thymocytes are stained. A subset of NK cells express this antigen weakly. The epitope recognized by this clone is sensitive to routine formalin fixation and paraffin embedding.
3rd Workshop: code no. 567

Immunogen

human thymocytes followed by peripheral blood T cells.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Flow cytometry/Cell sorting (1 paper)
Immunofluorescence (1 paper)
Monoclonal Anti-CD8 antibody produced in mouse is has been used for the following:
  • enumeration of total T cytotoxic/suppressor lymphocytes in bone marrow, blood and other body fluids
  • identification and localization of T cytotoxic/ suppressor lymphocytes in lymphoid and other tissues
  • analysis of cell mediated cytotoxicity
  • studies of immunoregulation in health and disease
  • investigation of natural killer (NK) cells
  • complement mediated cytolysis of CD8 expressing cells

Monoclonal Anti-CD8 antibody produced in mouse is suitable for flow cytometry at a dilution of 5μL using 1 × 106 cells.

Biochem/physiol Actions

CD8 might serve as a marker for neoplastic diseases and acute myeloid leukemia.
CD8a (Cluster of Differentiation 8a) is a human gene. CD8 binding to class I MHC and the co-receptor′s during the process of cytotoxic T-cell activation. T cells expresses T cell receptor (TCR) complexes which lacks CD3 delta, exhibit severely impaired positive selection and TCR-mediated activation of CD8 single-positive T cells. CD3 delta couples with TCR. The CD3 associated with CD8 is required for effective activation and positive selection of CD8(+) T cells. CD8 expression is a recurrent rare phenomenon in patients with chronic lymphocytic leukemia (CLL).

Target description

CD8 is a two chain complex (α/α, α/β) receptor for Class I MHC molecules. The receptor′s cytoplasmic portion is associated with p56lck protein tyrosine kinase which phosphorylates nearby proteins. The antigen is strongly expressed on approximately one third of mature T cells (cytotoxic/suppressor T cells).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kate Hsu et al.
Molecular cell, 14(2), 259-267 (2004-04-22)
Sequence analysis predicted significant structural homology between the HIV-1 accessory protein Vpu and the N-terminal region of TASK-1, a mammalian background K(+) channel. If the homology resulted from molecular piracy during HIV-1 evolution, these two proteins may have important functional
Saffron Dornan et al.
The Journal of biological chemistry, 277(3), 1912-1918 (2001-11-06)
An investigation into the role of CD45 isoforms in T cell antigen receptor signal transduction was carried out by transfecting CD45-negative CD4(+)CD8(+) HPB-ALL T cells with the CD45R0, CD45RBC, and CD45RABC isoforms. Fluorescence resonance energy transfer analysis showed that the
S Asselin et al.
European journal of immunology, 28(2), 532-539 (1998-04-01)
Polarization of T lymphocytes towards type 1 (T1) or type 2 (T2) subsets producing a distinct array of cytokines plays a role in several diseases and could be used for therapeutic intervention. Bearing this purpose in mind, we have established
An RFLP associated with pcDLeu2-14, a human T-cell differentiation antigen CD8 (Leu2) cDNA mapped to 2p12.
Bowcock AM, et al.
Nucleic Acids Research, 14(19), 7817-7817 (1986)
Kasturi Pal et al.
The Journal of cell biology, 212(7), 861-875 (2016-03-24)
Dynamic changes in membrane protein composition of the primary cilium are central to development and homeostasis, but we know little about mechanisms regulating membrane protein flux. Stimulation of the sonic hedgehog (Shh) pathway in vertebrates results in accumulation and activation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service