Skip to Content
Merck
All Photos(1)

Key Documents

C6116

Sigma-Aldrich

(+/-)-Cloprostenol sodium salt hydrate

≥98% (HPLC), powder

Synonym(s):

(+/-)-16-(3-Chlorophenoxy)-9,11,15R-trihydroxy-17,18,19,20-tetranorprosta-5Z,13E-dien-1-oic acid sodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H28ClNaO6 · xH2O
CAS Number:
Molecular Weight:
446.90 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

light brown

mp

68-70 °C

solubility

H2O: >10 mg/mL

storage temp.

−20°C

SMILES string

[Na+].[H]O[H].O[C@@H](COc1cccc(Cl)c1)\C=C\[C@H]2[C@H](O)C[C@H](O)[C@@H]2C\C=C/CCCC([O-])=O

InChI

1S/C22H29ClO6.Na.H2O/c23-15-6-5-7-17(12-15)29-14-16(24)10-11-19-18(20(25)13-21(19)26)8-3-1-2-4-9-22(27)28;;/h1,3,5-7,10-12,16,18-21,24-26H,2,4,8-9,13-14H2,(H,27,28);;1H2/q;+1;/p-1/b3-1-,11-10+;;/t16-,18-,19-,20+,21-;;/m1../s1

InChI key

ZEQHRVULQBYULU-LPFHDDPUSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Cloprostenol is a synthetic analogue of prostaglandin F2α (PGF). Cloprostenol contributes to the induction of estrus in animals and has uterotonic activity. Cloprostenol induces abortion in animals.

Application

(+/-)-Cloprostenol sodium salt hydrate has been used in studying its influence in oestrus synchronisation and uterine development in mice.

Biochem/physiol Actions

Cloprostenol is a potent FP prostanoid receptor agonist.
Cloprostenol is a potent FP prostanoid receptor agonist. It is 2- to 3-fold more potent than fluprostenol but less selective. Cloprostenol is similar in potency to PGF at EP and TP prostanoid receptors. It is used for the treatment of infertility in sows and gilts.

Features and Benefits

This compound is featured on the Prostanoid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New estrus synchronization and artificial insemination protocol for goats based on male exposure, progesterone and cloprostenol during the non-breeding season
Lopez-Sebastian A, et al.
Theriogenology, 68(8), 1081-1087 (2007)
Effect of progesterone on prostaglandin F2 alpha secretion and outcome of pregnancy during cloprostenol-induced abortion in mares.
Daels PF, et al.
American Journal of Veterinary Research, 57(9), 1331-1337 (1996)
Mano Sira et al.
Aesthetic surgery journal, 32(7), 822-824 (2012-09-04)
The prostaglandin F2a (PGF2a) analogue bimatoprost 0.03% (Allergan, Inc, Irvine, California) has been employed for the treatment of hypotrichosis since it gained Food and Drug Administration approval as Latisse in 2008. In this report, the authors retrospectively review the cases
X Valldecabres-Torres et al.
Journal of dairy science, 96(3), 1647-1652 (2013-01-15)
The 6-d timed artificial insemination protocol has been designed to advance luteolysis after the first administration of GnRH so that the preovulatory follicular diameter at second GnRH is reduced and thereby pregnancy outcome may be improved. To achieve an earlier
Muhammad Modu Bukar et al.
Reproductive biology, 12(3), 325-328 (2012-11-17)
The effects of estrus synchronization with prostaglandin F(2α) (PGF(2α)) and Controlled Internal Drug Release Device (CIDR) on ensuing antral follicular development were documented and compared to natural estrous cycles of non-seasonal tropical goats. Two to six follicular waves were observed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service