Skip to Content
Merck
All Photos(3)

Key Documents

C5678

Sigma-Aldrich

Carboxymethylcellulose sodium salt

low viscosity

Synonym(s):

Sodium carboxymethylcellulose

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

synthetic (organic)

form

powder

color

light yellow

useful pH range

(20 °C, 10 g/L, 68 °F neutral)

viscosity

50-200 cP, 4 % in H2O(25 °C)

mp

> 300.05  °C ((> 572.09 °F ))

solubility

water: soluble

cation traces

Na: 6.5-9.5%

storage temp.

room temp

InChI

1S/C6H12O6.C2H4O2.Na/c7-1-3(9)5(11)6(12)4(10)2-8;1-2(3)4;/h1,3-6,8-12H,2H2;1H3,(H,3,4);

InChI key

DPXJVFZANSGRMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is a low viscosity carboxymethylcellulose (CMC); the viscosity of a 4% solution in water at 25 oC is 50-200 centipoise (cps). The viscosity is both concentration and temperature dependent. As the temperature increases, the viscosity decreases. As the concentration increases, the viscosity increases. Low, medium and high viscosity carboxymethylcellulose (CMC) are all used as suspending agents. Low viscosity CMC is usually used in "thin" aqueous solutions. Medium viscosity CMC is used to make solutions that look like a syrup. High viscosity CMC is used to make a mixture, which resembles a cream or lotion.

Application

Soluble carboxymethylcelluloses (CM-cellulose; CMC) available in varying viscosities are used as viscosity modifiers (thickeners) to stabilize emulsions and as a chemical dispersants of oils and other carbon structures such as nanotubes. CMCs are used in the development of biostructures such as biofilms, emulsions and nanoparticles for drug delivery. Carboxymethylcellulose, low viscosity, may be used to make solutions the consistency of "thin" aqueous solutions.

Preparation Note

The product is soluble in water (40 mg/mL).The key to dissolving carboxymethylcellulose is to add the solid carefully to the water so that it is well dispersed (well-wetted). Adding the solid in portions may be necessary. Adding water to the dry solid produces a "clump" of solid that is very difficult to dissolve; the solid must be added to the water. Stir gently or shake intermittently; do not stir constantly with a magnetic stirring bar. High heat is not needed and may actually slow down the solubilization process. A mixing device, such as an impeller-type agitator which produces a vortex, would allow the powder to be drawn into the liquid, but it may produce some shearing.

Under normal conditions, the effect of temperature on solutions of this product is reversible, so slight temperature variation has no permanent effect on viscosity. However, long periods of heating CMC solutions at high temperatures (autoclaving) will degrade the product and permanently reduce viscosity. CMC is therefore very difficult to sterilize. γ-Irradiation, like heating, will degrade CMC. High viscosity CMC is more adversely affected by autoclaving and irradiation than is low viscosity CMC. Filtering CMC solutions tends to leave a gel behind because the material is fibrous, so solutions cannot be sterile filtered.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shigenobu Mitsuzawa et al.
Scientific reports, 7(1), 16320-16320 (2017-11-28)
The glycoside hydrolase family 7 (GH7) member cellobiohydrolase (CBH) is a key enzyme that degrades crystalline cellulose, an important structural component of plant cell walls. As GH7 CBH is a major component in the enzyme mixture used to degrade biomass
R Farenzena et al.
Journal of animal physiology and animal nutrition, 101(5), 1057-1066 (2016-07-28)
Two in vivo digestibility trials with sheep were conducted to identify the minimum period length of feeding a new diet to obtain reproducible values of nutritional variables onward and the minimum length of collection period as to obtain maximal precision for
C Philippeau et al.
Journal of dairy science, 100(4), 2637-2650 (2017-02-06)
This study investigated the effects of bacterial direct-fed microbials (DFM) on ruminal fermentation and microbial characteristics, methane (CH
Vinay Khatri et al.
Biotechnology for biofuels, 11, 144-144 (2018-05-26)
Pretreatment of lignocellulosic biomass (LCB) is a key step for its efficient bioconversion into ethanol. Determining the best pretreatment and its parameters requires monitoring its impacts on the biomass material. Here, we used fluorescent protein-tagged carbohydrate-binding modules method (FTCM)-depletion assay
Tim Wartewig et al.
Nature, 552(7683), 121-125 (2017-11-17)
T cell non-Hodgkin lymphomas are a heterogeneous group of highly aggressive malignancies with poor clinical outcomes. T cell lymphomas originate from peripheral T cells and are frequently characterized by genetic gain-of-function variants in T cell receptor (TCR) signalling molecules. Although

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service