Skip to Content
Merck
All Photos(2)

Key Documents

C4461

Sigma-Aldrich

Colistin sulfate salt

≥19,000 IU/mg

Synonym(s):

Polymyxin E

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
51102829
eCl@ss:
34050906
NACRES:
NA.85

form

powder

Quality Level

specific activity

≥19,000 IU/mg

antibiotic activity spectrum

Gram-negative bacteria

Mode of action

cell membrane | interferes

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(O)O.N1[C@H](C(=O)NCC[C@@H](C(=O)N[C@H](C(=O)N[C@@H](C(=O)N[C@H](C(=O)N[C@H](C(=O)N[C@H](C1=O)CCN)CCN)CC(C)C)CC(C)C)CCN)NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)CCCCC(CC)C)CCN)[C@H](O)C)CCN)[C@H](O)C

InChI

1S/C53H100N16O13.H2O4S/c1-9-30(6)12-10-11-13-41(72)60-33(14-20-54)48(77)69-43(32(8)71)53(82)65-36(17-23-57)45(74)64-38-19-25-59-52(81)42(31(7)70)68-49(78)37(18-24-58)62-44(73)34(15-21-55)63-50(79)39(26-28(2)3)67-51(80)40(27-29(4)5)66-46(75)35(16-22-56)61-47(38)76;1-5(2,3)4/h28-40,42-43,70-71H,9-27,54-58H2,1-8H3,(H,59,81)(H,60,72)(H,61,76)(H,62,73)(H,63,79)(H,64,74)(H,65,82)(H,66,75)(H,67,80)(H,68,78)(H,69,77);(H2,1,2,3,4)/t30?,31-,32-,33+,34+,35+,36+,37+,38+,39+,40-,42+,43+;/m1./s1

InChI key

ZJIWRHLZXQPFAD-LRYSGCCDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: peptide

Application

Colistin sulfate is used to permeabilize bacterial cell membranes and to study mannose-resistant haemagglutination and antibiotic resistance in certain organisms such as A. baumannii . It has been used to study hephrotoxicity in the rat kidney , and MICs, time-kill kinetics, and postantibiotic effect (PAE) against Pseudomonas aeruginosa .

Biochem/physiol Actions

Mode of Action: Binds to lipids on the cell cytoplasmic membrane of Gram-negative bacteria and disrupts the cell wall integrity.
Antimicrobial spectrum: Gram-negative bacteria.
Mode of Action: Binds to lipids on the cell cytoplasmic membrane of Gram-negative bacteria and disrupts the cell wall integrity.
Antimicrobial spectrum: Gram-negative bacteria. It is proposed that renal reabsorption of colistin may involve organic cation transporters and peptide transporters and that the process is sensitive to pH .

Analysis Note

Freely soluble in water, practically insoluble in acetone and in ethanol (96%)

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deepesh Nagarajan et al.
Science advances, 5(7), eaax1946-eaax1946 (2019-07-30)
Drug resistance is a public health concern that threatens to undermine decades of medical progress. ESKAPE pathogens cause most nosocomial infections, and are frequently resistant to carbapenem antibiotics, usually leaving tigecycline and colistin as the last treatment options. However, increasing
Ansley M Nemeth et al.
ChemMedChem, 15(2), 210-218 (2019-11-23)
Infections caused by multidrug-resistant (MDR) bacteria, particularly Gram-negative bacteria, are an escalating global health threat. Often clinicians are forced to administer the last-resort antibiotic colistin; however, colistin resistance is becoming increasingly prevalent, giving rise to the potential for a situation
Peng Cui et al.
Antimicrobial agents and chemotherapy, 60(11), 6867-6871 (2016-09-08)
Persisters are small populations of quiescent bacterial cells that survive exposure to bactericidal antibiotics and are responsible for many persistent infections and posttreatment relapses. However, little is known about how to effectively kill persister bacteria. In the work presented here
Mehri Haeili et al.
Microbial drug resistance (Larchmont, N.Y.), 24(9), 1271-1276 (2018-03-29)
Colistin is considered a last-hope antibiotic against extensively drug-resistant isolates of Acinetobacter baumannii. Resistance to colistin has been rarely reported for A. baumannii. Genetic alterations in the PmrA-PmrB two-component system and lipid A biosynthesis genes may be associated with colistin
Anna Ebbensgaard et al.
Frontiers in microbiology, 9, 2153-2153 (2018-09-25)
Bacterial resistance to classical antibiotics is emerging worldwide. The number of infections caused by multidrug resistant bacteria is increasing and becoming a serious threat for human health globally. In particular, Gram-negative pathogens including multidrug resistant Escherichia coli are of serious

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service