Skip to Content
Merck
All Photos(1)

Key Documents

C3556

Sigma-Aldrich

Catalase from human erythrocytes

≥90% (SDS-PAGE), buffered aqueous solution, ≥30,000 units/mg protein

Synonym(s):

H2O2:H2O2 oxidoreductase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

human erythrocytes

Quality Level

Assay

≥90% (SDS-PAGE)

form

buffered aqueous solution

specific activity

≥30,000 units/mg protein

mol wt

tetramer ~250 kDa

concentration

≤10 mg/mL

technique(s)

cell based assay: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C9H10O3/c1-2-12-9(11)7-3-5-8(10)6-4-7/h3-6,10H,2H2,1H3

InChI key

NUVBSKCKDOMJSU-UHFFFAOYSA-N

Gene Information

human ... CAT(847)

General description

Research area: Cell Signaling

Human catalase is a member of the monofunctional heme-containing catalases. It is an intracellular enzyme located at higher concentrations in the liver, erythrocytes, and kidney. Catalase is a homo-tetrameric protein and comprises amino acid residues, one heme group that is iron III protoporphyrin IX, and a nicotinamide adenine dinucleotide phosphate (NADPH) molecule. It is a ubiquitous enzyme found in most aerobic organisms. The catalase (CAT) gene is located on the human chromosome at 11p13.

Application

Catalase from human erythrocytes has been used:

  • to prevent reoxidation of reduced cytochrome c by H2O2 while measuring the production of superoxide with cytochrome C.
  • as a component of the imaging buffer for stochastic optical reconstruction microscopy (STORM) imaging of human skin fibroblasts
  • as a component of the gloxy mix for single-molecule imaging

Biochem/physiol Actions

Deficiency of catalase and oxidative stress are associated with vitiligo, hypertension, Alzheimer′s disease, diabetes mellitus, and acatalasemia. Catalase shows a dichotomous role in cancer as its expression is at a higher level in chronic lymphocytic leukemia, glioma, melanoma, and gastric cancer and lower levels of expression are seen in acute myeloid leukemia, pancreatic, lung, prostate, and skin (non-melanoma) cancers.
Catalase activates the decomposition of hydrogen peroxide, a reactive oxygen species, into water and oxygen. It functions as a natural antioxidant, protecting cells against oxidative damage to proteins, lipids and nucleic acids. Catalase has also been used to study the role reactive oxygen species play in gene expression and apoptosis.

Unit Definition

One unit will decompose 1.0 μmole of H2O2 per min at pH 7.0 at 25 °C, while the H2O2 conc. falls from 10.3 to 9.2 mM, measured by the rate of decrease of A240.

Physical form

Solution in 50 mM Tris, pH 8.0

Storage and Stability

Tightly closed. Dry. Keep locked up or in an area accessible only to qualified or authorized
persons

Analysis Note

Protein determined by biuret

inhibitor

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Urte Neniskyte et al.
Methods in molecular biology (Clifton, N.J.), 1041, 103-111 (2013-07-03)
Reactive oxygen and nitrogen species are both regulators and effectors of microglial activation, and assays of these oxidants can be used as a measure of acute and chronic activation of microglial cells. Here we describe quick methods to assess the
D A Chistiakov et al.
Diabetes/metabolism research and reviews, 20(3), 219-224 (2004-05-11)
Oxidative stress is involved in the origin of type 1 diabetes. Low efficiency of the scavenging antioxidant system has been shown to be related to the pathogenesis of the disease. This, therefore suggests that genes encoding catalase and other antioxidant
Investigation of the binding mechanism and inhibition of bovine liver catalase by quercetin: Multi-spectroscopic and computational study
Samaneh R, et al.
BioImpacts, 7(3), 147?153-147?153 (2017)
Li-Wen Shen et al.
Acta pharmacologica Sinica, 43(1), 177-193 (2021-07-24)
Inhibition of autophagy has been accepted as a promising therapeutic strategy in cancer, but its clinical application is hindered by lack of effective and specific autophagy inhibitors. We previously identified cepharanthine (CEP) as a novel autophagy inhibitor, which inhibited autophagy/mitophagy
Marilisa Galasso et al.
Free radical biology & medicine, 172, 264-272 (2021-06-16)
Aerobic organisms possess numerous antioxidant enzymatic families, including catalases, superoxide dismutases (SODs), peroxiredoxins (PRDXs), and glutathione peroxidases (GPXs), which work cooperatively to protect cells from an excess of reactive oxygen species (ROS) derived from endogenous metabolism or external microenvironment. Catalase

Articles

Cellular oxidative stress is countered by enzymatic scavengers and antioxidant modulators against reactive oxygen species damage.

Cellular oxidative stress is countered by enzymatic scavengers and antioxidant modulators against reactive oxygen species damage.

Cellular oxidative stress is countered by enzymatic scavengers and antioxidant modulators against reactive oxygen species damage.

Cellular oxidative stress is countered by enzymatic scavengers and antioxidant modulators against reactive oxygen species damage.

Protocols

This procedure may be used for all Catalase products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service