Skip to Content
Merck
All Photos(1)

Key Documents

C3230

Sigma-Aldrich

Coelenterazine h

solid

Synonym(s):

2-Deoxycoelenterazine, CLZN-h

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H21N3O2
CAS Number:
Molecular Weight:
407.46
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

form

solid

Quality Level

storage temp.

−20°C

SMILES string

Oc1ccc(cc1)C2=CN3C(=O)C(Cc4ccccc4)=NC3=C(Cc5ccccc5)N2

InChI

1S/C26H21N3O2/c30-21-13-11-20(12-14-21)24-17-29-25(22(27-24)15-18-7-3-1-4-8-18)28-23(26(29)31)16-19-9-5-2-6-10-19/h1-14,17,27,30H,15-16H2

InChI key

KAEGGIFPLJZUOZ-UHFFFAOYSA-N

General description

Coelenterazine is an imidazopyrazinone derivative. It is a substrate for bioluminescence in many luminous marine organisms.

Biochem/physiol Actions

10 times higher luminescence intensity than native coelenterazine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Liu et al.
Frontiers in plant science, 11, 583584-583584 (2020-12-18)
Cereal cyst nematodes are sedentary biotrophic endoparasites that secrete effector proteins into plant tissues to transit normal cells into specialized feeding sites and suppress plant defenses. To understand the function of nematode effectors in Heterodera avenae, here, we identified a
Yifan Zhang et al.
Scientific reports, 7(1), 7195-7195 (2017-08-05)
Epilepsy is a chronic brain disorder characterized by recurrent seizures due to abnormal, excessive and synchronous neuronal activities in the brain. It affects approximately 65 million people worldwide, one third of which are still estimated to suffer from refractory seizures.
Coelenterazine-dependent luciferases
Markova S V and Vysotski E S
Biochemistry (Moscow), 80(6), 714-732 (2015)
Lauren G Falkenberg et al.
Disease models & mechanisms, 14(5) (2021-05-25)
The pathology of primary ciliary dyskinesia (PCD) is predominantly attributed to impairment of motile cilia. However, PCD patients also have perplexing functional defects in myeloid cells, which lack motile cilia. Here, we show that coiled-coil domain-containing protein 103 (CCDC103), one
Md Alamgir Hossain et al.
Cell reports, 29(7), 1934-1945 (2019-11-14)
To evade immunity, many viruses express interferon antagonists that target STAT transcription factors as a major component of pathogenesis. Because of a lack of direct structural data, these interfaces are poorly understood. We report the structural analysis of full-length STAT1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service