Skip to Content
Merck
All Photos(7)

Key Documents

C2888

Sigma-Aldrich

Acetylcholinesterase from Electrophorus electricus (electric eel)

Type V-S, lyophilized powder, ≥1,000 units/mg protein

Synonym(s):

AChE, Acetylcholine acetylhydrolase, Cholinesterase, Acetyl

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

Electrophorus electricus

type

Type V-S

form

lyophilized powder

specific activity

≥1,000 units/mg protein

mol wt

280 kDa

composition

Protein, ≥45% biuret

pI 

~5.3

solubility

Tris buffer: 1 mg/mL (0.02 M Tris buffer, pH 7.5)

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Molecular Weight: 280 kDa
Isoelectric Point: 5.5
Extinction Coefficient: E1% = 18.0 (280 nm)

Acetylcholinesterase from Electrophorus electricus is a tetramer composed of 4 equal subunits of 70 kDa each. Each subunit contains one active site. The enzyme is a glycoprotein containing hexosamines.

Application

Acetylcholinesterase from Electrophorus electricus (electric eel) has been used:
  • in acetylcholinesterase activity assay[1]
  • to study its effects on Xenopus embryonic hippocampal cells[2]
  • to study the effect of acetylcholine inhibitors on development and plasticity[3]

Biochem/physiol Actions

Acetylcholinesterase (AChE) exhibits its activity against acetylcholine in the neuromuscular junctions and synaptic cleft of cholinergic neurons.[2] It participates in the nitric oxide signaling pathway.[1]
Major degradative enzyme for acetylcholine in vivo. Converts acetylcholine + H2O to choline + acetic acid.

Caution

For dilute solutions (<1 mg/ml), add 1 mg/ml BSA to stabilize. Solutions containing at least 1 mg protein/ml in dilute neutral phosphate buffer are stable at least 6 months refrigerated.

Unit Definition

One unit will hydrolyze 1.0 μmole of acetylcholine to choline and acetate per min at pH 8.0 at 37 °C.

Physical form

Lyophilized powder containing Tris buffer salts

Analysis Note

The activity obtained using acetylcholine as substrate is 30-100 times that obtained with butyrylcholine, using acetylcholinesterase from electric eel.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joana R Almeida et al.
Biomolecules, 10(8) (2020-08-06)
The development of alternative ecological and effective antifouling technologies is still challenging. Synthesis of nature-inspired compounds has been exploited, given the potential to assure commercial supplies of potential ecofriendly antifouling agents. In this direction, the antifouling activity of a series
Daniela Pereira et al.
Marine drugs, 19(1) (2020-12-31)
Over the last decades, antifouling coatings containing biocidal compounds as active ingredients were used to prevent biofouling, and eco-friendly alternatives are needed. Previous research from our group showed that polymethoxylated chalcones and glycosylated flavones obtained by synthesis displayed antifouling activity
Silvia Olivera et al.
Molecular and cellular neurosciences, 23(1), 96-106 (2003-06-12)
Here we show that chronic application of low concentrations (0.01-0.05 U/ml) or a single application of 1-5 U/ml acetylcholinesterase (AChE) promotes the extension of neuronal processes, synapse formation, and alpha-amino-3-hydroxy-5-methylisoxazolepropionate receptor (AMPAR) surface expression in both embryonic and postnatal hippocampal
Daniel A S Kitagawa et al.
Journal of enzyme inhibition and medicinal chemistry, 36(1), 1370-1377 (2021-06-22)
Organophosphorus poisoning caused by some pesticides and nerve agents is a life-threating condition that must be swiftly addressed to avoid casualties. Despite the availability of medical countermeasures, the clinically available compounds lack a broad spectrum, are not effective towards all
Nagham Alatrash et al.
ChemMedChem, 12(13), 1055-1069 (2017-06-13)
Four mononuclear [(L-L)2 Ru(tatpp)]2+ and two dinuclear [(L-L)2 Ru(tatpp)Ru(L-L)2 ]4+ ruthenium(II) polypyridyl complexes (RPCs) containing the 9,11,20,22-tetraazatetrapyrido[3,2-a:2',3'-c:3'',2''-l:2''',3'''-n]pentacene (tatpp) ligand were synthesized, in which L-L is a chelating diamine ligand such as 2,2'-bipyridine (bpy), 1,10-phenanthroline (phen), 3,4,7,8-tetramethyl-1,10-phenanthroline (Me4 phen) or 4,7-diphenyl-1,10-phenanthroline

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service