Skip to Content
Merck
All Photos(2)

Documents

C199

Sigma-Aldrich

CGS-15943

solid

Synonym(s):

9-Chloro-2-(2-furanyl)-[1,2,4]triazolo[1,5-c]quinazolin-5-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H8ClN5O
CAS Number:
Molecular Weight:
285.69
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

color

white

solubility

DMSO: >10 mg/mL
H2O: insoluble

storage temp.

room temp

SMILES string

Nc1nc2ccc(Cl)cc2c3nc(nn13)-c4ccco4

InChI

1S/C13H8ClN5O/c14-7-3-4-9-8(6-7)12-17-11(10-2-1-5-20-10)18-19(12)13(15)16-9/h1-6H,(H2,15,16)

InChI key

MSJODEOZODDVGW-UHFFFAOYSA-N

Application

CGS-15943 has been used as a non-selective adenosine receptor antagonist to study its effects on the proliferation of pancreatic ductal adenocarcinoma (PDAC) cells and hepatocellular carcinoma (HCC). It has also been used as a non-selective adenosine receptor antagonist to investigate the mechanism underlying adenosine inhibition on cholangiocarcinoma (CCA) cells.

Biochem/physiol Actions

CGS-15943 is a potent and non-selective adenosine receptor antagonist. It exhibits anti-carcinogenic and anti-apoptotic activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J m Li et al.
The Journal of surgical research, 80(2), 357-364 (1999-01-08)
Adenosine is a potent vasodilator of vascular smooth muscle. Endothelium-derived nitric oxide (NO) elicits vasodilation. We have previously reported that adenosine stimulates the production of NO from porcine carotid arterial endothelial cells (PCAEC) via a receptor-mediated mechanism. This study was
Elise M Weerts et al.
Psychopharmacology, 168(1-2), 155-163 (2003-04-02)
Caffeine and the adenosine A(1) and A(2A) receptor antagonist CGS15943 produce many behavioral effects that are similar to those produced by classic stimulant drugs (e.g. cocaine and amphetamines). The current study evaluated whether CGS15943 would maintain self-administration and reinstate extinguished
M Williams et al.
The Journal of pharmacology and experimental therapeutics, 241(2), 415-420 (1987-05-01)
CGS 15943A, a triazoloquinazoline, is a potent and selective adenosine receptor antagonist as assessed by its effects on radioligand binding and adenosine-stimulated adenylate cyclase activity in guinea pig synaptoneurosomes. At the adenosine A-1 receptor labeled with [3H]cyclohexyladenosine, CGS 15943A had
Lazaros C Foukas et al.
The Journal of biological chemistry, 277(40), 37124-37130 (2002-07-30)
We investigated the effects of methylxanthines on enzymatic activity of phosphoinositide 3-kinases (PI3Ks). We found that caffeine inhibits the in vitro lipid kinase of class I PI3Ks (IC(50) = 75 microm for p110 delta, 400 microm for p110 alpha and
A C Ngai et al.
American journal of physiology. Heart and circulatory physiology, 280(5), H2329-H2335 (2001-04-12)
The purpose of this study was to investigate the receptor subtypes that mediate the dilation of rat intracerebral arterioles elicited by adenosine. Penetrating arterioles were isolated from the rat brain, cannulated with the use of a micropipette system, and luminally

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service