Skip to Content
Merck
All Photos(1)

Documents

C0880

Sigma-Aldrich

Conalbumin from chicken egg white

Iron complex

Synonym(s):

Ovotransferrin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

biological source

chicken egg white

Quality Level

Assay

≥95% (agarose gel electrophoresis)

form

powder

technique(s)

gel permeation chromatography (GPC): suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

chicken ... TF(396241)

Looking for similar products? Visit Product Comparison Guide

General description

Iron-carrying proteins conalbumin and serum transferrin are synthesized with identical 19-amino-acid leader sequences that are removed during protein maturation. The leader sequences are presumably involved in secretion of the proteins by endoplasmic reticulum.

Application

Conalbumin (CA) is a monomeric glycoprotein that makes up approximately 13% of the albumin found in egg white. The iron complex of CA is more stable to chemical and physical treatments when compared to the uncomplexed proteins. At near neutral conditions, urea denaturation of CA modifies but does not destroy the iron complex.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The denaturation of proteins. IV. Conalbumin and iron(III)-conalbumin in urea solution.
A N GLAZER et al.
Biochimica et biophysica acta, 71, 109-123 (1963-04-02)
S N Thibodeau et al.
The Journal of biological chemistry, 253(11), 3771-3774 (1978-06-10)
The NH2-terminal sequences of egg white conalbumin and chicken serum transferrin were examined and found to be identical. Conalbumin, when synthesized in a rabbit reticulocyte cell-free translation system, was found to contain an NH2-terminal extension of 19 amino acid residues.
The resistances of conalbumin and its iron complex to physical and chemical treatments.
P R AZARI et al.
Archives of biochemistry and biophysics, 92, 44-52 (1961-01-01)
Alexandra Acero-Lopez et al.
Food chemistry, 135(4), 2245-2252 (2012-09-18)
High pressure processing of ovotransferrin was carried out to study the structural and physiochemical changes of ovotransferrin under various pressure levels. At pH 8 and pressures higher than 200 MPa, a decrease in total sulfhydryl groups and an increase in
Use of ovotransferrin on a turkey farm to reduce respiratory disease.
C Van Droogenbroeck et al.
The Veterinary record, 172(3), 71-71 (2012-12-25)

Articles

For use as a marker in SDS-PAGE; Albumin from chicken egg white, For use as a marker in SDS-PAGE; L-Lactic Dehydrogenase from rabbit muscle, Type XI, lyophilized powder, 600-1,200 units/mg protein

Chromatograms

application for HPLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service