BM991.8643.02
B Medical U201
Ultra Low Temperature Freezer
Synonym(s):
B Medical, Ultra Low Temperature Freezer
About This Item
Recommended Products
AC/DC input
220 V / 240 V, US 3-pin plug
material
stainless steel device (Coated Steel Ext Stainless Steel Int)
feature
CE compliant
CSA approved
packaging
pack of 1 ea
manufacturer/tradename
B Medical Systems
parameter
-20--86 °C temp. range
H × W × D
50.90 in. × 27.52 in. × 40.91 in.
capacity
7.55 ft3
weight
430 lb
shipped in
ambient
storage temp.
room temp
Looking for similar products? Visit Product Comparison Guide
General description
- Premium Upright, Ultra Low Temperature Freezer
- 1 ST Shelves
- 4 Casters with integrated feet
- 7" touchscreen Display
- One Insulated Inner Door
- Proximity Card Access Control
- RTMD Included
Application
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service