BAF188510016
Bel-Art® Freezer Storage Box
no. of holes, 100, assorted colors polypropylene, pack of 5
Synonym(s):
cryopreservation storage rack, cryovial storage, indexed tube rack, microtube storage
About This Item
Recommended Products
material
assorted colors polypropylene
feature
for tubes 0.5, 1.5, and 2.0 mL
no. of holes 100
packaging
pack of 5
manufacturer/tradename
Bel-Art F18851-0016
technique(s)
cryopreservation: suitable
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- Alphanumeric Indexing
- Translucent lid for easy access and viewing; matte panels can be used for labeling (100-place boxes have hinged lid, 81-place boxes have lift off lid)
- Withstand -80°C to +121°C (-112° to +249°F)
- Stackable with molded-in ′no slide′ features
- Assorted Colors with translucent lids (natural, blue, green, orange and purple)
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service