Skip to Content
Merck
All Photos(1)

Key Documents

B9655

Sigma-Aldrich

Anti-Avidin antibody, Mouse monoclonal, biotin conjugate

clone WC19.10, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-Avidin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

Quality Level

conjugate

biotin conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

WC19.10, monoclonal

form

buffered aqueous solution

technique(s)

direct ELISA: suitable
dot blot: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Avidin is a glycoprotein (68 kDa) composed of four identical polypeptides chains isolated from egg whites.Monoclonal Anti-Avidin (mouse IgG1 isotype) is derived from the hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse.
The antibody binds to the avidin molecule without interfering with the biotin-binding reaction. The product reacts specifically with egg white avidin, ExtrAvidin and their complexes formed with biotin. The antibody shows no cross-reaction with streptavidin.

Immunogen

Purified egg white avidin.

Application

Anti-Avidin-Biotin antibody, Mouse monoclonal has been used for fluorescence in situ hybridization (FISH) analysis of chromosomes X and 8 in lymphocytes isolated from women

Biochem/physiol Actions

Avidin has a high binding affinity for the vitamin, biotin. Biotin conjugated monoclonal anti-avidin increases specific avidin-biotin complex promoting the sensitivity of avidin-biotin immunoassays.

Physical form

Solution supplied in 0.01 M phosphate buffered saline, pH 7.4, containing 1% BSA and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chapter 16 - Biotechnological Approaches for Insect Pest Management
Integrated Pest Management Current Concepts and Ecological Perspective, 311-335 (2014)
Chapter 11 - (Strept)avidin?Biotin Systems
Bioconjugate Techniques, 465-505 (2013)
Jaeho Kim et al.
Ultrasonography (Seoul, Korea), 41(2), 382-393 (2021-12-29)
The purpose of this study was to evaluate if transcutaneous application of low-intensity ultrasound can locally enhance the effects of finasteride on hair growth in a murine model of androgenic alopecia (AA). AA mice (injected twice per week with testosterone
Murine monoclonal anti-avidin antibodies enhance the sensitivity of avidin-biotin immunoassays and immunohistologic staining
Cassano WF, et al.
Journal of Immunological Methods, 117(2), 169-174 (1989)
Spontaneous and spindle poison-induced micronuclei and chromosome non-disjunction in cytokinesis-blocked lymphocytes from two age groups of women
Bakou K, et al.
Mutagenesis, 17(3), 233-239 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service