Skip to Content
Merck
All Photos(1)

Documents

B9275

Sigma-Aldrich

Buprenorphine hydrochloride

Synonym(s):

[5α,7α(S)]-17-(Cyclopropylmethyl)-α-(1,1-dimethylethyl)4,5-epoxy-18,19-dihydo-3-hydroxy-6-methoxy-α-methyl-6,14-ethenomorphinan-7-methanol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H41NO4 · HCl
CAS Number:
Molecular Weight:
504.10
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

Quality Level

drug control

USDEA Schedule III; Home Office Schedule 3; psychotrope (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IIC (Portugal)

SMILES string

Cl[H].CO[C@]12CC[C@@]3(C[C@@H]1[C@](C)(O)C(C)(C)C)[C@H]4Cc5ccc(O)c6O[C@@H]2[C@]3(CCN4CC7CC7)c56

InChI

1S/C29H41NO4.ClH/c1-25(2,3)26(4,32)20-15-27-10-11-29(20,33-5)24-28(27)12-13-30(16-17-6-7-17)21(27)14-18-8-9-19(31)23(34-24)22(18)28;/h8-9,17,20-21,24,31-32H,6-7,10-16H2,1-5H3;1H/t20-,21-,24-,26+,27-,28+,29-;/m1./s1

InChI key

UAIXRPCCYXNJMQ-RZIPZOSSSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Full agonist at ORL1 receptors, antagonist or partial agonist at μ, κ, and δ opioid receptors.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soumen Chakraborty et al.
Journal of medicinal chemistry, 64(18), 13873-13892 (2021-09-11)
Mitragynine and 7-hydroxymitragynine (7OH) are the major alkaloids mediating the biological actions of the psychoactive plant kratom. To investigate the structure-activity relationships of mitragynine/7OH templates, we diversified the aromatic ring of the indole at the C9, C10, and C12 positions
Maria Hauge Pedersen et al.
The Journal of biological chemistry, 296, 100503-100503 (2021-03-09)
G protein-coupled receptors (GPCRs) signal through activation of G proteins and subsequent modulation of downstream effectors. More recently, signaling mediated by β-arrestin has also been implicated in important physiological functions. This has led to great interest in the identification of
Lino Becerra et al.
The Journal of pharmacology and experimental therapeutics, 345(1), 41-51 (2013-02-02)
Pharmacological magnetic resonance imaging (phMRI) is one method by which a drug's pharmacodynamic effects in the brain can be assessed. Although phMRI has been frequently used in preclinical and clinical settings, the extent to which a phMRI signature for a
Alexander S Hauser et al.
Cell, 172(1-2), 41-54 (2017-12-19)
Natural genetic variation in the human genome is a cause of individual differences in responses to medications and is an underappreciated burden on public health. Although 108 G-protein-coupled receptors (GPCRs) are the targets of 475 (∼34%) Food and Drug Administration
Hisham Alhaddad et al.
Toxicology, 305, 99-108 (2013-02-05)
While most deaths from asphyxia related to buprenorphine (BUP) overdose have been reported in males, higher plasma concentrations of BUP and its toxic metabolite norbuprenorphine (NBUP) have been observed in females. We previously demonstrated that P-glycoprotein (P-gp) modulation at the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service