Skip to Content
Merck
All Photos(1)

Key Documents

B8563

Sigma-Aldrich

Aniline Blue solution

2.5% in 2% acetic acid

Synonym(s):

Acid blue 93; Cl 42780

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H25N3O9S3Na2
CAS Number:
Molecular Weight:
737.73
MDL number:
UNSPSC Code:
41116121
NACRES:
NA.47

form

liquid

concentration

2.5% in 2% acetic acid

technique(s)

microbe id | staining: suitable

color

dark blue

solubility

water: soluble

λmax

0.21-0.37 at 597-603 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

N(c2ccc(cc2)C(=C4C=CC(=Nc5ccccc5)C=C4)c3cc(c(cc3)N)C)c1ccccc1

InChI

1S/C32H27N3/c1-23-22-26(16-21-31(23)33)32(24-12-17-29(18-13-24)34-27-8-4-2-5-9-27)25-14-19-30(20-15-25)35-28-10-6-3-7-11-28/h2-22,34H,33H2,1H3

InChI key

LBLFVSANTXZPEX-UHFFFAOYSA-N

General description

Aniline Blue is the traditional name applied to anionic dyes produced by successive phenylation and sulfonation of basic fuchsine. Aniline blue consists of sulfonation products of variable mixtures of phenylated rosaniline and pararosaniline. It is a mixture of methyl blue and water blue. The dye is also known as cotton blue, and ink blue. Methyl blue of aniline blue belongs to the group of triaminotriphenylmethane dyes.

Application

Aniline blue (also known as cotton blue and ink blue) is a component of commonly used polychrome stains. It is used to stain collagen fibers in tissue sections using Masson′s trichrome protocol for staining multiple components. Collagen is stained blue by this method. The dye is suitable for selective staining of callose in plant specimens and staining histones for assessing nuclear maturity. Heidenhain′s AZAN stain and the Mallory′s polychrome stain both entail staining steps using aniline blue-orange G mixtures. Aniline blue is used in Gomori′s one-step trichrome stain and Mallory′s connective tissue stain for tissue including kidney and intestine.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of membrane proteome of Paracoccidioides lutzii and its regulation by zinc
Juliana Santana de Curcio et al.
Future science OA (2017)
Lilian C Baeza et al.
Frontiers in microbiology, 8, 2308-2308 (2017-12-13)
The genus
Lan-Tao Gou et al.
Cell, 180(6), 1212-1227 (2020-03-15)
The paternal genome undergoes a massive exchange of histone with protamine for compaction into sperm during spermiogenesis. Upon fertilization, this process is potently reversed, which is essential for parental genome reprogramming and subsequent activation; however, it remains poorly understood how this
Rafal P Piprek et al.
Cells, 8(12) (2019-12-15)
Normal gonad development assures the fertility of the individual. The properly functioning gonads must contain a sufficient number of the viable germ cells, possess a correct architecture and tissue structure, and assure the proper hormonal regulation. This is achieved by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service