Skip to Content
Merck
All Photos(2)

Documents

B8049

Sigma-Aldrich

5-Bromo-4-chloro-3-indolyl β-D-glucuronide cyclohexylammonium salt

tablet

Synonym(s):

X-GlcA, X-glucuronide CHA salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H13BrClNO7 · C6H13N
CAS Number:
Molecular Weight:
521.79
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

form

tablet

Quality Level

storage temp.

−20°C

SMILES string

NC1CCCCC1.O[C@@H]2[C@@H](O)[C@@H](O[C@@H]([C@H]2O)C(O)=O)Oc3c[nH]c4ccc(Br)c(Cl)c34

InChI

1S/C14H13BrClNO7.C6H13N/c15-4-1-2-5-7(8(4)16)6(3-17-5)23-14-11(20)9(18)10(19)12(24-14)13(21)22;7-6-4-2-1-3-5-6/h1-3,9-12,14,17-20H,(H,21,22);6H,1-5,7H2/t9-,10-,11+,12-,14+;/m0./s1

InChI key

JXCKZXHCJOVIAV-CYRSAHDMSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5-Bromo-4-chloro-3-indolyl β-D-glucuronide cyclohexylammonium salt has been used as a histochemical reagent in X-GLUC staining.
Chromogenic substrate for β-glucuronidase (GUS) gene detection.

Biochem/physiol Actions

5-bromo-4-chloro-3-indolyl-β-D-glucuronide sodium salt (X-GlcA) is a colorless substrate. The pre-immobilized X-GlcA can be converted by β-glucuronidase enzyme, synthesised by Escherichia coli.

Quantity

Contains 10 mg substrate per tablet.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service