Skip to Content
Merck
All Photos(1)

Documents

B6404

Supelco

BCIP®/NBT solution, premixed

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105333
NACRES:
NA.32

form

liquid

Quality Level

Application

BCIP/NBT solution, premixed has been used:
  • as a substrate to develop dot-blots
  • as a substrate to reveal alkaline phosphatase in the immunohistochemical analysis
  • to detect specific immune complexes in western blot analysis

Other Notes

0.48 mM NBT, 0.56 mM BCIP, 10 mM Tris and 59.3 mM MgCl2, pH approx. 9.2

Substrates

Substrate for visualizing alkaline phosphatase conjugates in Western blotting.

Legal Information

BCIP is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Slow-and fast-twitch rat hind limb skeletal muscle phenotypes 8 months after spinal cord transection and olfactory ensheathing glia transplantation
Negredo P, et al.
The Journal of Physiology, 586(10), 2593-2610 (2008)
Neuroprotective effects of recombinant human granulocyte colony-stimulating factor (G-CSF) in neurodegeneration after optic nerve crush in rats
Tsai R K, et al.
Experimental Eye Research, 87(3), 242-250 (2008)
Willie Lin et al.
Oncotarget, 9(2), 1992-2001 (2018-02-09)
CD34 is a transmembrane phosphoglycoprotein used to selectively enrich bone marrow in hematopoietic stem cells for transplantation. Treating rats with CD34
Dietary iron concentration may influence aging process by altering oxidative stress in tissues of adult rats
Arruda L F, et al.
PLoS ONE, 8(4), e61058-e61058 (2013)
The major human structural IgE epitope of the Brazil nut allergen Ber e 1: a chimaeric and protein microarray approach
Alcocer M J C, et al.
Journal of Molecular Biology, 343(3), 759-769 (2004)

Articles

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service