Skip to Content
Merck
All Photos(2)

Documents

B4936

Sigma-Aldrich

Brassicasterol

from semisynthetic

Synonym(s):

5,22-Cholestadien-24β-methyl-3β-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H46O
CAS Number:
Molecular Weight:
398.66
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.77

biological source

semisynthetic

Quality Level

Assay

≥98% (TLC)

form

powder

shipped in

wet ice

storage temp.

2-8°C

SMILES string

CC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C28H46O/c1-18(2)19(3)7-8-20(4)24-11-12-25-23-10-9-21-17-22(29)13-15-27(21,5)26(23)14-16-28(24,25)6/h7-9,18-20,22-26,29H,10-17H2,1-6H3/b8-7+/t19-,20+,22-,23-,24+,25-,26-,27-,28+/m0/s1

InChI key

OILXMJHPFNGGTO-ZAUYPBDWSA-N

Related Categories

Application

Brassicasterol was used as standard to analyze the sterols from mussels sample by thin layer chromatography.

Biochem/physiol Actions

Brassicasterol is a plant sterol, structurally similar to cholesterol. Plant sterols compete with cholesterol and reduce the content of cholesterol incorporated into micelles thereby reducing its absorption. Brassicasterol reportedly decreases the progression of atherosclerosis. The level of brassicasterol in cerebrospinal fluid may be used as a prognostic factor for progression of Alzheimer′s disease.
Brassicasterol is a phytosterol found in rapeseed and canola oils; it is also present in marine algae and shellfish. Brassicasterol has been shown to inhibit sterol Δ24-reductase, an enzyme involved in the mammalian cholesterol biosynthesis pathway.

Preparation Note

Brassicasterol is from a semi-synthetic source. It yields clear, colorless solution in chloroform at 50 mg/ml.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

T Vanmierlo et al.
Acta psychiatrica Scandinavica, 124(3), 184-192 (2011-05-19)
Plant sterols (sitosterol, campesterol, stigmasterol and brassicasterol) are solely dietary-derivable sterols that are structurally very similar to cholesterol. In contrast to peripheral cholesterol, plant sterols can cross the blood-brain barrier and accumulate within mammalian brain. As an impaired function of
Gavin Tansley et al.
Current gerontology and geriatrics research, 2012, 179318-179318 (2012-06-01)
Over the past 15 years, insights into sterol metabolism have improved our understanding of the relationship between lipids and common conditions such as atherosclerosis and Alzheimer's Disease (AD). A better understanding of sterol lipid metabolism in individuals with Down Syndrome
The search of new biomarkers to identify Alzheimer's disease: an editorial comment to T. Vanmierlo et al. 'The plant sterol brassicasterol and additional CFS biomarker in Alzheimer's Disease' (1).
A Lobo et al.
Acta psychiatrica Scandinavica, 124(3), 163-164 (2011-05-14)
Karen J Murphy et al.
Asia Pacific journal of clinical nutrition, 12(1), 50-60 (2003-05-10)
In view of previously reported anti-inflammatory bioactivity of the New Zealand Green Lipped Mussel (NZGLM), the overall lipid profile and fatty acid and sterol composition of the NZGLM from various sites in New Zealand (Hallam Cove, Port Ligar. Little Nikau)
E Heggen et al.
Nutrition, metabolism, and cardiovascular diseases : NMCD, 20(4), 258-265 (2009-09-15)
Data comparing the impact of different sources of plant sterols on CVD risk factors and antioxidant levels is scarce. We evaluated the effects of plant sterols from rapeseed and tall oils on serum lipids, lipoproteins, fat-soluble vitamins and plant sterol

Articles

Cholesterol synthesis regulation by dietary levels, LDL receptors control lipid-rich LDL particle transport in cells.

Cholesterol synthesis regulation by dietary levels, LDL receptors control lipid-rich LDL particle transport in cells.

Cholesterol synthesis regulation by dietary levels, LDL receptors control lipid-rich LDL particle transport in cells.

Cholesterol synthesis regulation by dietary levels, LDL receptors control lipid-rich LDL particle transport in cells.

Protocols

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service