Skip to Content
Merck
All Photos(1)

Key Documents

B4061

Sigma-Aldrich

BATCP

≥98% (HPLC), solid

Synonym(s):

(S)-[5-Acetylamino-1-(2-oxo-4-trifluoromethyl-2H-chromen-7-ylcarbamoyl)pentyl]carbamic acid tert-butyl ester, Cpd 3b

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H28F3N3O6
CAS Number:
Molecular Weight:
499.48
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

solubility

DMSO: 16 mg/mL

storage temp.

2-8°C

SMILES string

CC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(=O)Nc1ccc2c(OC(=O)C=C2C(F)(F)F)c1

InChI

1S/C23H28F3N3O6/c1-13(30)27-10-6-5-7-17(29-21(33)35-22(2,3)4)20(32)28-14-8-9-15-16(23(24,25)26)12-19(31)34-18(15)11-14/h8-9,11-12,17H,5-7,10H2,1-4H3,(H,27,30)(H,28,32)(H,29,33)/t17-/m0/s1

InChI key

NVKNRJCOVHAZDK-KRWDZBQOSA-N

Related Categories

Biochem/physiol Actions

HDAC 6 selective substrate (over HDAC 1, Class II over Class I).

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service