Skip to Content
Merck
All Photos(1)

Key Documents

AV48999

Sigma-Aldrich

Anti-NEK7 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-NIMA (never in mitosis gene a)-related kinase 7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

34 kDa

species reactivity

rat, human, mouse, dog, guinea pig, bovine, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NEK7(140609)

Immunogen

Synthetic peptide directed towards the N terminal region of human NEK7

Application

Anti-NEK7antibody produced in rabbit is suitable for western blotting at a concentration of 1.0μg/ml.

Biochem/physiol Actions

NIMA-related kinase 7 (NEK7), a serine/threonine protein kinase is required for the proper formation of spindle during mitosis. NEK7 recruits centrosomal pericentriolar material (PCM) proteins which are required for centriole duplication and spindle pole formation. Optimum activity of NEK7 is necessary for cell cycle progression during M and G1 phase and during cytokinesis.

Sequence

Synthetic peptide located within the following region: KARADCIKEIDLLKQLNHPNVIKYYASFIEDNELNIVLELADAGDLSRMI

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nek9, Nek6, Nek7 and the separation of centrosomes.
Sara Sdelci et al.
Cell cycle (Georgetown, Tex.), 10(22), 3816-3817 (2011-11-09)
Sunghwan Kim et al.
Journal of cell science, 124(Pt 22), 3760-3770 (2011-11-22)
The centrosomes in dividing cells follow a series of cyclical events of duplication and separation, which are tightly linked to the cell cycle. Serine/threonine-protein kinase NEK7 (NEK7) is a centrosomal kinase that is required for proper spindle formation during mitosis.
Laura O'Regan et al.
Molecular and cellular biology, 29(14), 3975-3990 (2009-05-06)
Nek6 and Nek7 are members of the NIMA-related serine/threonine kinase family. Previous work showed that they contribute to mitotic progression downstream of another NIMA-related kinase, Nek9, although the roles of these different kinases remain to be defined. Here, we carried

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service