Skip to Content
Merck
All Photos(2)

Documents

AV32533

Sigma-Aldrich

Anti-SPDEF (AB1) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Spdef Antibody, Spdef Antibody - Anti-SPDEF (AB1) antibody produced in rabbit, Anti-PDEF, Anti-RP11-375E1__A.3, Anti-SAM pointed domain containing ets transcription factor, Anti-bA375E1.3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

37 kDa

species reactivity

bovine, rabbit, human, dog, guinea pig

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SPDEF(25803)

Related Categories

General description

Rabbit polyclonal anti-SPDEF (AB1) antibody reacts with human, mouse, rat, bovine, pig, and canine SAM pointed domain containing ets transcription factors.
SAM pointed domain containing ets transcription factor (SPDEF, PDEF) is involved in the regulation of terminal differentiation of goblet/Paneth progenitor cells into intestinal Paneth and goblet cells. SPDEF plays a critical role in regulating a transcriptional network mediating IL-13-induced MUC5AC synthesis dependent on STAT6.
SPDEF suppresses the metastasis of prostate cancer and modulates goblet cell hyperplasia in airway epithelial cells.
Rabbit Anti-SPDEF (AB1) antibody recognizes human, mouse, rat, bovine, pig, and canine SPDEF.

Immunogen

Synthetic peptide directed towards the middle region of human SPDEF

Application

Rabbit Anti-SPDEF (AB1) antibody can be used for western blot applications at a concentration of 1μg/ml.
Rabbit polyclonal anti-SPDEF (AB1) antibody is used to tag SAM pointed domain containing ets transcription factor for detection and quantitation by immunocytochemical and immunohistochemical (IHC) techniques. It is used as a probe to determine the presence and roles of SAM pointed domain containing ets transcription factor in the differentiation of Paneth and goblet cells and mucin production.

Biochem/physiol Actions

PDEF is an ETS transcription factor expressed in prostate epithelial cells. It acts as an androgen-independent transactivator of PSA expression.PDEF is an ETS transcription factor expressed in prostate epithelial cells. It acts as an androgen-independent transactivator of PSA (MIM 176820) expression.[supplied by OMIM]. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

Sequence

Synthetic peptide located within the following region: ELCAMSEEQFRQRSPLGGDVLHAHLDIWKSAAWMKERTSPGAIHYCASTS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joshua J Steffan et al.
The Journal of biological chemistry, 287(35), 29968-29978 (2012-07-05)
Emerging evidence suggests that the SAM pointed domain containing ETS transcription factor (SPDEF) plays a significant role in tumorigenesis in prostate, breast, colon, and ovarian cancer. However, there are no in vivo studies with respect to the role of SPDEF
Kwon-Sik Park et al.
The Journal of clinical investigation, 117(4), 978-988 (2007-03-10)
Goblet cell hyperplasia and mucous hypersecretion contribute to the pathogenesis of chronic pulmonary diseases including cystic fibrosis, asthma, and chronic obstructive pulmonary disease. In the present work, mouse SAM pointed domain-containing ETS transcription factor (SPDEF) mRNA and protein were detected

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service