Skip to Content
Merck
All Photos(2)

Key Documents

A8681

Sigma-Aldrich

3′-Sialyllactose

from bovine milk or colostrum, ≥97% (HPLC)

Synonym(s):

α-NeuNAc-(2→3)-β-D-Gal-(1→4)-D-Glc, 3′-N-Acetylneuraminyl-D-lactose sodium salt, 3′-SL, 3′-Sialyl-D-lactose, NANA-Lactose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H39NO19
CAS Number:
Molecular Weight:
633.55
Beilstein:
75357
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

bovine milk or colostrum

Quality Level

Assay

≥97% (HPLC)

form

powder

color

white

solubility

water: 20 mg/mL, clear, colorless

cation traces

Na: 3.1-4.7%

storage temp.

−20°C

SMILES string

[Na+].[H]C(=O)[C@H](O)[C@@H](O)[C@H](O[C@@H]1O[C@H](CO)[C@H](O)[C@H](O[C@@]2(C[C@H](O)[C@@H](NC(C)=O)[C@@H](O2)[C@H](O)[C@H](O)CO)C([O-])=O)[C@H]1O)[C@H](O)CO

InChI

1S/C23H39NO19.Na/c1-7(29)24-13-8(30)2-23(22(38)39,42-19(13)15(35)10(32)4-26)43-20-16(36)12(6-28)40-21(17(20)37)41-18(11(33)5-27)14(34)9(31)3-25;/h3,8-21,26-28,30-37H,2,4-6H2,1H3,(H,24,29)(H,38,39);/q;+1/p-1/t8-,9-,10+,11+,12+,13+,14+,15+,16-,17+,18+,19+,20-,21-,23-;/m0./s1

InChI key

LTWFUJWFLMHANB-TZCPRLTCSA-M

Looking for similar products? Visit Product Comparison Guide

Application

3′-Sialyllactose is a compound where in the acetylneuraminyl (NANA) unit is connected to the galactosyl unit of lactose at the 3 position. In 6′-sialylactose, this connection is at the 6 position. 3′-Sialyllactose and 6′-Sialyllactose are used to differentiate and characterize binding domains of viruses, such as influenza and rhinitis viruses, that recognize NANA capped cell surface receptors. 3′-Sialyllactose and 6′-sialylactose are used as reference compounds in analytical methods developed to measure their presence in materials such as milk and colostrums.

Other Notes

To gain a comprehensive understanding of our extensive range of Oligosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J V Mysore et al.
Gastroenterology, 117(6), 1316-1325 (1999-12-02)
Helicobacter pylori can be eradicated by administration of antimicrobials, but resistant strains have emerged, and there is a need for novel therapeutic approaches against this infection. This study aimed to determine the safety and efficacy of 3'-sialyllactose sodium salt (3'SL)
Maarten H Wilbrink et al.
Journal of agricultural and food chemistry, 63(25), 5976-5984 (2015-06-06)
Decoration of prebiotic galacto-oligosaccharides (GOS) with sialic acid yields mixtures of GOS and sialylated GOS (Sia-GOS), novel products that are expected to have both prebiotic and antiadhesive functionalities. The recombinantly produced trans-sialidase enzyme from Trypanosoma cruzi (TcTS), an enzyme with
S Crennell et al.
Nature structural biology, 7(11), 1068-1074 (2000-11-04)
Paramyxoviruses are the main cause of respiratory disease in children. One of two viral surface glycoproteins, the hemagglutinin-neuraminidase (HN), has several functions in addition to being the major surface antigen that induces neutralizing antibodies. Here we present the crystal structures
T Nakamura et al.
Carbohydrate research, 329(2), 471-476 (2000-12-16)
The composition of the products formed by treatment of commercial alpha-Neu5Ac-(2 --> 3)-beta-D-Galp-(1 --> 4)-D-Glc (3'-sialyllactose) with glacial acetic acid was investigated by 1H-13C one- and two-dimensional NMR spectroscopy and fast atom bombardment-mass spectrometry. The data confirmed that the major
Oriane Machon et al.
Glycobiology, 27(2), 123-128 (2016-11-01)
Glycosylation is a group of post-translational modifications that displays a large variety of structures and are implicated in a plethora of biological processes. Therefore, studying glycosylation requires different technical approaches and reliable tools, lectins being part of them. Here, we

Articles

O-Linked glycoproteins are usually large proteins with a molecular mass of >200 kDa. Glycosylation generally occurs in high-density clusters and may represent as much as 50-80% of the overall mass.

O-Linked glycoproteins are usually large proteins with a molecular mass of >200 kDa. Glycosylation generally occurs in high-density clusters and may represent as much as 50-80% of the overall mass.

O-Linked glycoproteins are usually large proteins with a molecular mass of >200 kDa. Glycosylation generally occurs in high-density clusters and may represent as much as 50-80% of the overall mass.

O-Linked glycoproteins are usually large proteins with a molecular mass of >200 kDa. Glycosylation generally occurs in high-density clusters and may represent as much as 50-80% of the overall mass.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service