Skip to Content
Merck
All Photos(4)

Key Documents

A8001

Sigma-Aldrich

Aconitine

≥95% (HPLC), crystalline

Synonym(s):

Acetylbenzoylaconine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H47NO11
CAS Number:
Molecular Weight:
645.74
Beilstein:
74608
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

crystalline

color

white

solubility

H2O: 0.3 mg/mL
ethanol: 35 mg/mL

SMILES string

CCN1C[C@]2(COC)[C@H](O)C[C@@H](OC)C34C5C[C@]6(O)[C@@H](OC)[C@H](O)[C@@](OC(C)=O)(C5[C@H]6OC(=O)c7ccccc7)C([C@H](OC)C23)C14

InChI

1S/C34H47NO11/c1-7-35-15-31(16-41-3)20(37)13-21(42-4)33-19-14-32(40)28(45-30(39)18-11-9-8-10-12-18)22(19)34(46-17(2)36,27(38)29(32)44-6)23(26(33)35)24(43-5)25(31)33/h8-12,19-29,37-38,40H,7,13-16H2,1-6H3/t19-,20-,21-,22-,23+,24+,25?,26?,27+,28-,29+,31+,32-,33?,34-/m1/s1

InChI key

XFSBVAOIAHNAPC-VBUFWTEXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Aconitine has been used:
  • to study its cardiotoxic effects along the pericardium meridian (PM) on cardiac rhythm in rabbits
  • as a standard in high-performance thin layer chromatography (HPTLC) fingerprinting method
  • in the aconitine-based lipo-alkaloids semi-synthesis

Aconitine is a neurotoxin which activates tetrodotoxin-sensitive Na+ channels, inducing presynaptic depolarization and blocking the release of neurotransmitters. Aconitine also blocks norepinephrine reuptake. In the heart, aconitine induces ventricular tachycardia after intracoronary injection.

Biochem/physiol Actions

Aconitine is a diesterditerpene alkaloid found abundantly in the plant Aconitum genera. It possesses analgesic, antipyretic and antirheumatic activity. Aconitine is involved in blocking neurotransmission. It acts as a neurotoxin as well as a cardiotoxin. Aconitine triggers ventricular tachycardia (VT) and ventricular fibrillation (VF). It interacts with voltage-dependent Na+ channels which results in the depolarization of membranes. Aconitine may exhibit therapeutic effects against systemic lupus erythematosus (SLE). It also inhibits the reuptake of norepinephrine.
Neurotoxin. Activates tetrodotoxin-sensitive Na+ channels, inducing presynaptic depolarization, thus blocking the nerve action potential which, in turn, blocks the release of neurotransmitters and decreases the end plate potential at the neuromuscular junction. Aconitine also blocks norepinephrine reuptake. In the heart, aconitine induces ventricular tachycardia after intracoronary injection. In cultured ventricular myocytes, aconitine increases the duration of the action potential and induces the appearance of early after depolarization.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Oral - Acute Tox. 2 Inhalation

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Peijian Tong et al.
Journal of ethnopharmacology, 146(2), 562-571 (2013-02-05)
Fuzi (lateral root of Aconitum carmichaeli) is a popular traditional Chinese medicine well known for its both therapeutic and high-toxic activities. Its toxic alkaloid ingredients, mainly aconitine, mesaconitine, and hypaconitine, are responsible for the high toxicity. However, to date, no
Ling Ye et al.
Toxicology letters, 216(2-3), 86-99 (2012-12-04)
Aconitum alkaloids including aconitine (AC), mesaconitine (MA), hypaconitine (HA), are highly toxic. Their hydrolysates, such as benzoylaconine (BAC), benzoylmesaconine (BMA), benzoylhypaconine (BHA), aconine, and mesaconine, are considerably less toxic. Efflux transporters, including P-glycoprotein (P-gp), breast cancer resistance protein (BCRP), and
Bio-analytical studies on the process of detoxification and safety evaluation of Aconitum laciniatum and Abrus precatorius for use in ayurvedic preparations
Bapat, S P and Sane, R T
International Journal of Pharmaceutical Sciences and Research, 3(3), 914-914 (2012)
A Ameri
Progress in neurobiology, 56(2), 211-235 (1998-10-07)
Preparations of Aconitum roots are employed in Chinese and Japanese medicine for analgesic, antirheumatic and neurological indications. The recent surge in use of phytomedicine derived from traditional Chinese medicine as well as increasing concerns about possible toxic effects of these
Hongbin Zhu et al.
Talanta, 103, 56-65 (2012-12-04)
A fingerprinting approach was developed by means of UPLC-ESI/MS(n) (ultra-performance liquid chromatography-electrospray ionization/mass spectrometry) for the quality control of processed Radix Aconiti, a widely used toxic traditional herbal medicine. The present fingerprinting approach was based on the two processing methods

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service