Skip to Content
Merck
All Photos(1)

Key Documents

A7642

Supelco

Albumin from chicken egg white

For use as a marker in SDS-PAGE

Synonym(s):

Albumin from hen egg white, Ovalbumin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41105337
eCl@ss:
42010201
NACRES:
NA.25

biological source

chicken egg white

Quality Level

form

powder

mol wt

44287 Da by calculation
~45 kDa

packaging

vial of 25 mg

technique(s)

electrophoresis: suitable

UniProt accession no.

storage temp.

2-8°C

Gene Information

chicken ... OVAL(396058)

Looking for similar products? Visit Product Comparison Guide

Application

A phosphorylated glycoprotein which is useful as a marker (~45 kDa) for protein electrophoresis. Has been used to produce an animal model of asthma.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongwei Han et al.
The Journal of clinical investigation, 124(12), 5442-5452 (2014-11-05)
Atopic dermatitis (AD) and food allergy are closely linked; however, the mechanisms that guide the progression of AD to allergic inflammatory responses at other mucosal surfaces, including the gastrointestinal tract, are not well understood. Here, we determined that exposure of
Alexandre Panchaud et al.
Analytical chemistry, 83(6), 2250-2257 (2011-02-24)
Data-dependent precursor ion selection is widely used in shotgun proteomics to profile the protein components of complex samples. Although very popular, this bottom-up method presents major drawbacks in terms of detectable dynamic range. Recently, we demonstrated the superior performance of
Hongwei Han et al.
Journal of immunology (Baltimore, Md. : 1950), 190(3), 904-912 (2013-01-01)
The epithelial-derived cytokine thymic stromal lymphopoietin (TSLP) has been associated with the promotion of type 2 inflammation and the induction of allergic disease. In humans TSLP is elevated in the lungs of asthma patients and in the lesional skin of
John A Gebe et al.
Matrix biology plus, 9, 100052-100052 (2021-03-16)
The extracellular matrix glycosaminoglycan hyaluronan (HA) accumulates in human and mouse islets during the onset of autoimmune type 1 diabetes (T1D). HA plays a critical role in T1D pathogenesis, as spontaneous disease is blocked in mice fed the HA synthesis
Hongwei Han et al.
JCI insight, 8(3) (2023-01-11)
Ocular surface diseases, including conjunctivitis, are recognized as common comorbidities in atopic dermatitis (AD) and occur at an increased frequency in patients with AD treated with biologics targeting IL-4 receptor α (IL-4Rα) or IL-13. However, the inflammatory mechanisms underlying this

Articles

For use as a marker in SDS-PAGE; Albumin from chicken egg white, For use as a marker in SDS-PAGE; L-Lactic Dehydrogenase from rabbit muscle, Type XI, lyophilized powder, 600-1,200 units/mg protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service