Skip to Content
Merck
All Photos(1)

Key Documents

A7512

Sigma-Aldrich

N-Acetyl-DL-phenylalanine β-naphthyl ester

≥98% (TLC), suitable for ligand binding assays

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H19NO3
CAS Number:
Molecular Weight:
333.38
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

N-Acetyl-DL-phenylalanine β-naphthyl ester,

Assay

≥98% (TLC)

form

powder

technique(s)

ligand binding assay: suitable

color

white

storage temp.

2-8°C

SMILES string

CC(=O)NC(Cc1ccccc1)C(=O)Oc2ccc3ccccc3c2

InChI

1S/C21H19NO3/c1-15(23)22-20(13-16-7-3-2-4-8-16)21(24)25-19-12-11-17-9-5-6-10-18(17)14-19/h2-12,14,20H,13H2,1H3,(H,22,23)

InChI key

BBXRRTJNJCPGBU-UHFFFAOYSA-N

Biochem/physiol Actions

N-Acetyl-DL-phenylalanine β-naphthyl ester (NAPBNE), a chromogenic substrate, is used to identify, differentiate and characterize serine protease(s) and peptidase(s).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Y Darani et al.
Parasitology, 115 ( Pt 3), 237-247 (1997-09-23)
A cationic Schistosoma mansoni cercarial antigen was shown to be a serine protease as it was capable of hydrolysing N-acetyl-DL-phenylalanine beta-naphthyl ester (NAPBNE) after precipitation by immunoelectrophoresis, and this reaction was modulated by the serine protease inhibitors phenylmethanesulfonyl fluoride (PMSF)
M A Jongsma et al.
Analytical biochemistry, 212(1), 79-84 (1993-07-01)
An improved, time efficient, visual assay for quantitative determination of proteinase inhibitor activity in protein extracts is reported. Proteinase inhibitor activity of mammalian, bacterial, and fungal serine proteinases can be quantified. The method relies on radial diffusion of proteinase inhibitor
P Collin-Osdoby et al.
Molecular & general genetics : MGG, 243(6), 674-680 (1994-06-15)
Mutations at the apeA locus in Salmonella typhimurium lead to loss of a soluble enzyme ("protease I") that hydrolyzes the chromogenic endoprotease substrate N-acetyl phenylalanine beta-naphthyl ester. We have isolated pseudorevertants of S. typhimurium apeA mutations that have regained the
H Y Darani et al.
Parasitology, 135(4), 467-472 (2008-01-23)
An enzyme found previously in extracts of adult Schistosoma mansoni worms, that hydrolysed the chromogenic substrate N-acetyl-DL-phenylalanine beta-naphthyl-ester, has here been further investigated and characterized. Evidence that the molecule found in the parasite was antigenically and enzymatically homologous with a
K Havemann et al.
Klinische Wochenschrift, 61(1), 49-56 (1983-01-03)
Two cytochemical methods for detection of granulocytic elastase and chymotrypsin employing alanine and phenylalanine naphthyl esters were developed. Specificity of reaction with the ester substrates was proven by chloromethyl ketone inhibitors. The results of both staining methods were almost identical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service