Skip to Content
Merck
All Photos(1)

Key Documents

A7111

Sigma-Aldrich

Acepromazine maleate

≥98% (HPLC)

Synonym(s):

Acetopromazine maleate salt, 2-Acetyl-10-(3-dimethylaminopropyl)phenothiazine maleate salt, Acepromazine maleate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22N2OS · C4H4O4
CAS Number:
Molecular Weight:
442.53
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

yellow

solubility

H2O: >10 mg/mL

storage temp.

room temp

SMILES string

OC(=O)\C=C/C(O)=O.CN(C)CCCN1c2ccccc2Sc3ccc(cc13)C(C)=O

InChI

1S/C19H22N2OS.C4H4O4/c1-14(22)15-9-10-19-17(13-15)21(12-6-11-20(2)3)16-7-4-5-8-18(16)23-19;5-3(6)1-2-4(7)8/h4-5,7-10,13H,6,11-12H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;2-1-

InChI key

FQRHOOHLUYHMGG-BTJKTKAUSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Acepromazine maleate has been used:
  • as a component of anesthesia cocktail to test its effects on behavioral changes in rats
  • as a component of anesthesia cocktail to study its effects on lung resistance of rats
  • as an anti-psychotic pharmaceutical agent to study its cytotoxic effects on malignant glioma in glioblastoma (GBM) cells

Biochem/physiol Actions

Acepromazine is a phenothiazine antipsychotic comonly used as a veterinary drug (horses, dogs and cats). The compound is an analogue antipsychotic drug chlorpromazine (#C8138). The drug is thought to block receptors of dopamine in the brain. Recently it was discover that the compound inhibits ABCG2 transported protein, which overexpression in tumors is associated with drug resistance.
Atypical neuroleptic.

Features and Benefits

This compound is featured on the Dopamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Age-Dependent Progression in Lung Pathophysiology can be Prevented by Restoring Fatty Acid and Ceramide Imbalance in Cystic Fibrosis
Youssef M, et al.
Lung, 13(3), 1-11 (2020)
Akari Kamine et al.
The Japanese journal of veterinary research, 60(1), 5-13 (2012-03-31)
The metabolic mechanisms to circannual changes in body mass of bears have yet to be elucidated. We hypothesized that the Japanese black bear (Ursus thibetanus japonicus) has a metabolic mechanism that efficiently converts carbohydrates into body fat by altering insulin
Effect of hypoxia-inducible factor-1/vascular endothelial growth factor signaling pathway on spinal cord injury in rats
Chen H, et al.
Experimental and Therapeutic Medicine, 13(3), 861-866 (2017)
Bobbi J Conner et al.
American journal of veterinary research, 73(5), 595-601 (2012-04-27)
To evaluate the effect of acepromazine maleate administered IV on platelet function assessed in healthy dogs by use of a modified thromboelastography assay. 6 healthy adult mixed-breed dogs. Dogs received each of 3 treatments (saline [0.9% NaCl] solution [1 to
J M McGree et al.
Journal of veterinary pharmacology and therapeutics, 36(1), 31-42 (2013-01-16)
We describe the population pharmacokinetics of an acepromazine (ACP) metabolite (2-(1-hydroxyethyl)promazine) (HEPS) in horses for the estimation of likely detection times in plasma and urine. ACP (30 mg) was administered to 12 horses, and blood and urine samples were taken

Articles

We offer many products related to dopamine receptors for your research needs.

We offer many products related to dopamine receptors for your research needs.

We offer many products related to dopamine receptors for your research needs.

We offer many products related to dopamine receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service