Skip to Content
Merck
All Photos(1)

Key Documents

A6667

Sigma-Aldrich

Anti-Rabbit IgG (whole molecule), F(ab′)2 fragment−Peroxidase antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.54

biological source

goat

conjugate

peroxidase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

direct ELISA: 1:10,000

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Related Categories

General description

IgG antibodies regulate several functions such as complement activation and phagocytosis. Thus they play a crucial role in facilitating cytological immune responses. Anti-rabbit IgG (whole molecule), F(ab′)2 fragment-peroxidase antibody can be used to detect immune-complexes. The product can also be used in direct ELISA. Goat anti- rabbit IgG (whole molecule), F(ab′)2 fragment-peroxidase antibody reacts specifically with all rabbit immunoglobulins.

Immunogen

Purified rabbit IgG.

Application

Anti-rabbit IgG (whole molecule), F(ab′)2 fragment-peroxidase antibody can be used in immunoprecipitation and western blotting.
Proteins from lysed Leishmania amazonensis cells were analyzed by western blot using HRP-conjugated goat anti-rabbit Fc specific IgG as the secondary antibody.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin with preservative.

Preparation Note

Prepared using the periodate method described by Wilson, M.B., and Nakane, P.K., in Immunofluorescence and Related Staining Techniques, Elsevier/North Holland Biomedical Press, Amsterdam, p215 (1978).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qi Feng et al.
Experimental and therapeutic medicine, 15(3), 2597-2603 (2018-02-20)
Stromal cell-derived factor-1 (SDF-1) predicts poor clinical outcomes of certain types of cancer. Furthermore, vascular endothelial growth factor (VEGF) promotes the growth and metastasis of solid tumors. The aim of the present study was to examine the expression of SDF-1
Krishan Kumar et al.
JCI insight, 2(3), e88032-e88032 (2017-02-15)
The fibrotic reaction, which can account for over 70%-80% of the tumor mass, is a characteristic feature of human pancreatic ductal adenocarcinoma (PDAC) tumors. It is associated with activation and proliferation of pancreatic stellate cells (PSCs), which are key regulators
Jae Hoon Jeong et al.
Tuberculosis and respiratory diseases, 80(3), 247-254 (2017-07-28)
Airway epithelial cells are the first line of defense, against pathogens and environmental pollutants, in the lungs. Cellular stress by cadmium (Cd), resulting in airway inflammation, is assumed to be directly involved in tissue injury, linked to the development of
María P Iturriaga et al.
Oncology letters, 14(2), 2198-2206 (2017-08-07)
Cyclooxygenase (COX)-2 expression is positively correlated with malignant features in canine mammary carcinomas. Non-steroidal anti-inflammatory drugs (NSAIDs) inhibit COX activity and may therefore possess anticancer effects. Meloxicam is an NSAID that is widely used in human and veterinary medicine. High
E M Benkhart et al.
Journal of immunology (Baltimore, Md. : 1950), 165(3), 1612-1617 (2000-07-21)
IL-10 is a unique cytokine because it is anti-inflammatory and immunosuppressive. IL-10 is regulated at the level of transcription, but the critical motifs and the relevant transcription factors controlling this gene have remained elusive to date. We now report that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service