Skip to Content
Merck
All Photos(1)

Key Documents

A5862

Sigma-Aldrich

Anisomycin, Ready Made Solution from Streptomyces griseolus

10 mg/mL in DMSO

Synonym(s):

(2R,3S,4S)-2-(4-Methoxybenzyl)-3,4-pyrrolidinediol-3-acetate, 2-[(4-Methoxyphenyl)methyl]-3,4-pyrrolidinediol 3-acetate, Flagecidin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H19NO4
CAS Number:
Molecular Weight:
265.30
EC Number:
MDL number:
UNSPSC Code:
51102829
NACRES:
NA.85

form

liquid

Quality Level

concentration

10 mg/mL in DMSO

antibiotic activity spectrum

Gram-negative bacteria

Mode of action

protein synthesis | inhibits

shipped in

dry ice

storage temp.

−20°C

SMILES string

N1[C@H]([C@H]([C@@H](C1)O)OC(=O)C)Cc2ccc(cc2)OC

InChI

1S/C14H19NO4/c1-9(16)19-14-12(15-8-13(14)17)7-10-3-5-11(18-2)6-4-10/h3-6,12-15,17H,7-8H2,1-2H3/t12-,13+,14+/m0/s1

InChI key

YKJYKKNCCRKFSL-BFHYXJOUSA-N

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K L Shires et al.
Nature communications, 3, 1246-1246 (2012-12-06)
In isolated hippocampal slices, decaying long-term potentiation can be stabilized and converted to late long-term potentiation lasting many hours, by prior or subsequent strong high-frequency tetanization of an independent input to a common population of neurons-a phenomenon known as 'synaptic
Einar Ö Einarsson et al.
Learning & memory (Cold Spring Harbor, N.Y.), 19(10), 449-452 (2012-09-18)
It has been suggested that memories become more stable and less susceptible to the disruption of reconsolidation over weeks after learning. Here, we test this by targeting the anterior cingulate cortex (ACC) and test its involvement in the formation, consolidation
Ayumi Kishioka et al.
Molecular brain, 6, 17-17 (2013-04-17)
Fear is one of the most potent emotional experiences and is an adaptive component of response to potentially threatening stimuli. Cumulative evidence suggests that the amygdala plays a central role in the acquisition, storage and expression of fear memory. We
Louis C Leung et al.
Nature neuroscience, 16(2), 166-173 (2013-01-08)
Cell adhesion molecules and diffusible cues both regulate axon pathfinding, yet how these two modes of signaling interact is poorly understood. The homophilic cell adhesion molecule NF-protocadherin (NFPC) is expressed in the mid-dorsal optic tract neuroepithelium and in the axons
Yalan Zhang et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 32(44), 15318-15327 (2012-11-02)
Loss of the RNA-binding protein fragile X mental retardation protein (FMRP) represents the most common form of inherited intellectual disability. Studies with heterologous expression systems indicate that FMRP interacts directly with Slack Na(+)-activated K(+) channels (K(Na)), producing an enhancement of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service