Skip to Content
Merck
All Photos(1)

Key Documents

A5187

Sigma-Aldrich

Anti-Sheep IgG (whole molecule)–Alkaline Phosphatase antibody produced in donkey

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Donkey Anti-Sheep IgG (whole molecule)–AP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

donkey

Quality Level

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

technique(s)

direct ELISA: 1:30,000
western blot: 1:30,000

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Affinity isolated antibody is obtained from anti-sheep IgG antiserum by immunospecific purification which removes essentially all donkey serum proteins, including immunoglobulins, which do not specifically bind to sheep IgG. Anti-Sheep IgG is conjugated to alkaline phosphatase by protein cross-linking with 0.2% glutaraldehyde. Specificity of the antiserum is determined by immunoelectrophoresis (IEP), prior to conjugation. By IEP, the antiserum reacts specifically with normal sheep serum and sheep IgG. Identity and purity of the antibody is established by immunoelectrophoresis, prior to conjugation. Electrophoresis of the product followed by diffusion against anti-horse IgG and anti-donkey whole serum results in single arcs of precipitation.
IgGs are glycoprotein antibodies that modulate several immune responses. Sheep IgGs against target proteins are often used as primary antibodies in various research applications. Thus, secondary anti-sheep IgGs are useful tools for the analysis of target proteins. Anti-Sheep IgG (whole molecule)-Alkaline Phosphatase antibody binds to all sheep immunoglobulins.

Specificity

normal sheep serum and sheep IgG

Immunogen

Purified sheep IgG

Application

Anti-Sheep IgG (whole molecule)-Alkaline Phosphatase antibody is suitable for use in ELISA .
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)
Solid-Phase Decapeptide Assays were performed using alkaline phosphatase conjugated donkey anti-sheep IgG at 1:1000 for 2-3 hours at room temperature.
The anibody may be used in applications such as immunoblotting, dot blot and direct ELISA at a dilution of 1:30,000

Physical form

Solution in 0.05 M Tris, pH 8.0, containing 1 mM MgCl2, 50% glycerol, 1% bovine serum albumin, and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael C DeSantis et al.
The Journal of biological chemistry, 291(25), 13088-13097 (2016-04-30)
The envelope glycoprotein (Env) gp120/gp41 is required for HIV-1 infection of host cells. Although in general it has been perceived that more Env gives rise to higher infectivity, the precise quantitative dependence of HIV-1 virion infectivity on Env density has
Transfer conductance in second growth Douglas-fir (Pseudotsuga menziesii (Mirb.)Franco) canopies
Warren, C., R., et al
Plant, Cell and Environment, 26(8), 1215-1227 (2003)
Kirk M Brown et al.
Nature biotechnology, 40(10), 1500-1508 (2022-06-03)
Therapeutics based on short interfering RNAs (siRNAs) delivered to hepatocytes have been approved, but new delivery solutions are needed to target additional organs. Here we show that conjugation of 2'-O-hexadecyl (C16) to siRNAs enables safe, potent and durable silencing in
C Z Alvarado et al.
Poultry science, 83(6), 1039-1046 (2004-06-23)
Pale, soft, and exudative (PSE) meat is a growing problem in the turkey industry and has been associated with processing conditions such as slow carcass chilling. The development of PSE meat is caused by protein denaturation resulting from a rapid
György Szláma et al.
The FEBS journal, 280(16), 3822-3839 (2013-07-09)
Myostatin, a negative regulator of skeletal muscle growth, is produced from myostatin precursor by multiple steps of proteolytic processing. After cleavage by a furin-type protease, the propeptide and growth factor domains remain associated, forming a noncovalent complex, the latent myostatin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service