Skip to Content
Merck
All Photos(1)

Key Documents

A4847

Sigma-Aldrich

Alsterpaullone

≥98% (HPLC), powder

Synonym(s):

9-Nitro-7,12-dihydroindolo-[3,2-d][1]benzazepin-6(5)-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H11N3O3
CAS Number:
Molecular Weight:
293.28
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

yellow to brown

solubility

DMSO: ≥10 mg/mL

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1ccc2[nH]c-3c(CC(=O)Nc4ccccc-34)c2c1

InChI

1S/C16H11N3O3/c20-15-8-12-11-7-9(19(21)22)5-6-14(11)18-16(12)10-3-1-2-4-13(10)17-15/h1-7,18H,8H2,(H,17,20)

InChI key

OLUKILHGKRVDCT-UHFFFAOYSA-N

Gene Information

Application

Alsterpaullone has been used to stabilize β-catenin.
Alsterpaullone inhibits GSK-3β, CDK5/p25, and CDK1/cyclin B. Alsterpaullone has also been studied as an antimanic therapeutic for the acute manic phase of bipolar disorder.

Biochem/physiol Actions

Alsterpaullone (ALP) is a cyclin-dependent kinase (CDK) inhibitor. It is considered as a therapeutic agent for group 3 medulloblastomas. Alsterpaullone modulates the progression of cell cycle. It can arrest cell cycle and stimulate the apoptosis of cancer cells by activating caspase-9 through perturbation of mitochondrial membrane potential.
Potent and selective inhibitor of GSK-3β and CDK5/p25; potent inhibitor of CDK1/cyclin B (IC50 = 0.035 μM).

Features and Benefits

This compound is featured on the CDKs and GSK-3 pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mikhail Kalinichev et al.
The international journal of neuropsychopharmacology, 14(8), 1051-1067 (2011-01-07)
There is a growing body of evidence suggesting that animal models can be developed to probe the specific domains of bipolar disorder (BD) using the endophenotype approach. Here we tested clinically active antimanic drugs to validate amphetamine-induced hyperactivity in Black
C Schultz et al.
Journal of medicinal chemistry, 42(15), 2909-2919 (1999-07-30)
The paullones represent a novel class of small molecule cyclin-dependent kinase (CDK) inhibitors. To investigate structure-activity relationships and to develop paullones with antitumor activity, derivatives of the lead structure kenpaullone (9-bromo-7,12-dihydroindolo[3,2-d][1]benzazepin-6(5H)-one, 4a) were synthesized. Paullones with different substituents in the
Michael Trevino et al.
Developmental dynamics : an official publication of the American Association of Anatomists, 240(12), 2673-2679 (2011-11-05)
Although regeneration is widespread among metazoa, the molecular mechanisms have been studied in only a handful of taxa. Of these taxa, fewer still are amenable to studies of embryogenesis. Our understanding of the evolution of regeneration, and its relation to
Tsuneo Takadera et al.
Neurotoxicity research, 21(3), 309-316 (2011-10-19)
In the central nervous system, members of the Src family of tyrosine kinases (SFKs) are widely expressed and are abundant in neurons. The purpose of this study is to examine whether glycogen synthase-3 (GSK-3) is involved in SFK inhibitor-induced apoptosis.
Laura Iglesias Ollé et al.
Biomedicines, 12(6) (2024-06-27)
Hydra head formation depends on an organizing center in which Wnt/β-catenin signaling, that plays an inductive role, positively regulates Sp5 and Zic4, with Sp5 limiting Wnt3/β-catenin expression and Zic4 triggering tentacle formation. Using transgenic lines in which the HySp5 promoter

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service