Skip to Content
Merck
All Photos(1)

Documents

A4656

Sigma-Aldrich

Anti-Mouse IgG (whole molecule)−Alkaline Phosphatase antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Goat Anti-Mouse IgG (whole molecule)−AP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse

should not react with

human

technique(s)

direct ELISA: 1:1,000

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Immunoglobulin G (IgG) is a glycoprotein antibody that regulates immune responses such as phagocytosis and is also involved in the development of autoimmune diseases. Mouse IgGs have four distinct isotypes, namely, IgG1, IgG2a, IgG2b, and IgG3. IgG1 regulates complement fixation in mice.
Goat Anti-Mouse IgG (whole molecule)-Alkaline Phosphatase antibody binds to mouse IgGs and does not cross-react with human serum proteins.

Immunogen

purified mouse IgG

Application

Goat Anti-Mouse IgG (whole molecule)-Alkaline Phosphatase antibody has been used for ELISA assays.
Immunoprecipitated 293T cells lysates were analyzed by western blot using alkaline phosphatase conjugated goat anti-mouse IgG as the secondary antibody at 1:7500.
Surfactant Protein A was detected in bronchoalveolar fluid using alkaline phosphatase conjugated goat anti-mouse IgG as the secondary at μg/ml in TBS/Tween containing final concentration of 0.5M NaCl.

Other Notes

Antibody adsorbed with human serum proteins.

Physical form

Solution in 0.05 M Tris, pH 8.0, containing 1% bovine serum albumin, 1 mM MgCl2 and 15 mM sodium azide.

Preparation Note

Adsorbed to reduce background with human samples.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carlos Andrés Rodriguez-Salazar et al.
Viruses, 14(2) (2022-02-27)
Dengue virus is a ssRNA+ flavivirus, which produces the dengue disease in humans. Currently, no specific treatment exists. siRNAs regulate gene expression and have been used systematically to silence viral genomes; however, they require controlled release. Liposomes show favorable results
The LIM-only Protein DRAL/FHL2 Interacts with and Is a Corepressor for the Promyelocytic Leukemia Zinc Finger Protein
McLoughlin, P.
The Journal of Cell Biology, 277, 37045-37053 (2002)
Delia Piedad Recalde-Reyes et al.
Peptides, 153, 170797-170797 (2022-04-05)
Dengue virus (DENV) infection is mediated by the interaction between the virus envelope protein and cellular receptors of the host cells. In this study, we designed peptides to inhibit protein-protein interaction between dengue virus and CD44 receptor, which is one
P J Simpson et al.
Circulation, 81(1), 226-237 (1990-01-01)
Pentobarbital anesthetized dogs were subjected to 90 minutes of left circumflex coronary artery (LCCA) occlusion followed by 72 hours of reperfusion. Control or anti-Mo1 (904) F(ab')2 fragments of monoclonal antibodies were administered intravenously at a dose of 1 mg/kg beginning
Julia Knöckel et al.
Molecular & cellular proteomics : MCP, 20, 100038-100038 (2021-01-31)
Sporozoites are a motile form of malaria-causing Plasmodium falciparum parasites that migrate from the site of transmission in the dermis through the bloodstream to invade hepatocytes. Sporozoites interact with many cells within the host, but the molecular identity of these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service