Skip to Content
Merck

A4596

ANTI-FLAG® M1 Agarose Affinity Gel

Synonym(s):

Anti-ddddk, Anti-dykddddk

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.32
UNSPSC Code:
12352203
Conjugate:
agarose conjugate
Species reactivity:
-
Application:
Citations:
81
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


conjugate

agarose conjugate

Quality Segment

antibody product type

primary antibodies

form

suspension

isotype

IgG12b

capacity

≥0.6 mg/mL, gel binding capacity

storage temp.

−20°C

General description

ANTI-FLAG® M1 Agarose Affinity Gel is a purified IgG2B monoclonal antibody covalently attached to agarose.

Application

For purification of N-terminal FLAG fusion proteins. Since binding is Ca2+-dependent, proteins can be eluted with a buffer containing EDTA, as well as by the standard methods using either FLAG peptide or glycine-HCl buffer, pH 3. ANTI-FLAG M1 does not bind to Met-FLAG fusion proteins, so this resin is not appropriate for purifying unprocessed, cytoplasmically expressed fusion proteins.

Affinity gel is for calcium mediated purification of N-terminal FLAG fusion proteins.

immunoprecipitation (IP): suitable

Elution - FLAG peptide, Glycine, pH 3.5 EDTA

Learn more product details in our FLAG® application portal.

Biochem/physiol Actions

Binding specificity: Free N-Terminus of FLAG sequence
N-Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys-C

Features and Benefits

  • Typically purify fusion proteins from crude lysates to single band purity in just one chromatography step.
  • Fusion protein may be eluted from affinity resin by mild elution with EDTA.
  • A solution of FLAG peptide can be used for gentle, non-denaturing elution of FLAG fusion proteins.

Physical form

Suspension of beaded agarose in 50% glycerol containing 10 mM sodium phosphate, 150 mM NaCl, pH 7.4, 0.02% (w/v) sodium azide

Other Notes

ANTI-FLAG® M1 does not bind to Met-FLAG fusion proteins, so this resin is not appropriate for purifying unprocessed, cytoplasmically expressed fusion proteins.

Legal Information

ANTI-FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library