A4021
Nε-Acetyl-L-lysine
≥98% (TLC)
Synonym(s):
N6-acetyl-L-lysine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3CONH(CH2)4CH(NH2)CO2H
CAS Number:
Molecular Weight:
188.22
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26
Recommended Products
Product Name
Nε-Acetyl-L-lysine,
Assay
≥98% (TLC)
Quality Level
form
powder
concentration
50 mg/mL in 80% acetic acid
color
colorless to white
mp
250 °C (dec.) (lit.)
storage temp.
−20°C
SMILES string
CC(=O)NCCCC[C@H](N)C(O)=O
InChI
1S/C8H16N2O3/c1-6(11)10-5-3-2-4-7(9)8(12)13/h7H,2-5,9H2,1H3,(H,10,11)(H,12,13)/t7-/m0/s1
InChI key
DTERQYGMUDWYAZ-ZETCQYMHSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Nε-Acetyl L-α Lysine Improves Activity and Stability of α-Amylase at Acidic Conditions: A Comparative Study with other Osmolytes. This study highlights the use of Nε-Acetyl-ʟ-lysine in enhancing the functional stability and activity of α-amylase under acidic conditions, demonstrating its potential as a valuable additive in industrial enzyme applications (Joghee et al., 2020).
Biochem/physiol Actions
Nε-Acetyl-L-lysine (L-AcK) is an R-chain N-acetylated α amino acid used together with other lysine analogues to differentiate and characterized various aminoacylases and regulator 2 (Sir2) enzymes/sirtuins.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Morten B Trelle et al.
Analytical chemistry, 80(9), 3422-3430 (2008-03-15)
Tandem mass spectrometry (MS/MS) is a powerful tool for characterization of post-translationally modified proteins, including epsilon-N-acetyllysine-containing species. Previous reports indicate that epsilon-N-acetyllysine immonium ions are useful marker ions for peptides containing epsilon-N-acetyllysine, but the specificity and sensitivity of these ions
Ying Huang et al.
Molecular bioSystems, 6(4), 683-686 (2010-03-20)
By overexpressing the C-terminal domain of the ribosomal protein L11 to decrease release factor 1-mediated termination of protein translation, enhanced amber suppression is achieved in E. coli. This enhanced amber suppression efficiency allows the genetic incorporation of three N(epsilon)-acetyl-l-lysines into
K Hoffmann et al.
Die Pharmazie, 55(8), 601-606 (2000-09-16)
Inhibitors of histone deacetylase (HD) are of great potential as new drugs due to their ability to influence transcriptional regulation and to induce apoptosis or differentiation in cancer cells. So far only radioactive enzyme activity assays or in-vivo assays with
A Pähler et al.
Chemical research in toxicology, 11(9), 995-1004 (1998-10-07)
Antibodies directed against chemical specific protein modifications are valuable tools to detect and comparatively quantify protein modifications. Both Nepsilon-(dichloroacetyl)-L-lysine and Nepsilon-(trichloroacety)l-L-lysine have been detected as modified amino acids in liver and kidneys of rats treated with perchloroethene (PER) after proteolysis.
T Henle et al.
Zeitschrift fur Lebensmittel-Untersuchung und -Forschung, 198(1), 66-67 (1994-01-01)
After heating N alpha-acetyllysine and glucose for 4 h at 90 degrees C in the dry state and subsequent acid hydrolysis with 7.8 N HCl, preparative fractionation of the dihydrochlorides of furosine and pyridosine was achieved by cation-exchange chromatography. The
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service