Skip to Content
Merck
All Photos(2)

Key Documents

A3981

Sigma-Aldrich

Anti-β-Amyloid antibody, Mouse monoclonal

enhanced validation

clone BAM-10, purified from hybridoma cell culture

Synonym(s):

Anti-A-BETA, Anti-Amyloid β Precursor Protein, Clone BAM91

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

BAM-10, monoclonal

form

buffered aqueous solution

species reactivity

human

packaging

antibody small pack of 25 μL

enhanced validation

independent
Learn more about Antibody Enhanced Validation

concentration

~1.5 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 2.5-5 μg/mL using formic-acid treated tissue sections of human Alzheimer disease brain tissue
indirect ELISA: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... APP(351) , APP(351)

Looking for similar products? Visit Product Comparison Guide

General description

β-amyloid gene is located on human chromosome 21q21.
The β-amyloid precursor protein (APP) is cleaved sequentially by the proteolytic enzymes β-and γ-secretase to produce β-amyloid (Aβ) peptides with the Aβ1-42 and the Aβ1-40 forms being the most prevalent. Secreted Aβ peptides are degraded either via a re-uptake mechanism followed by endosomal degradation, or by an extracellular insulin degrading enzyme. Extracellular accumulation of Aβ leads to the formation of aggregates, fibrils and eventually amyloid deposits called neuritic plaques, which is the hallmark of Alzheimer′s disease (AD).
The antibody reacts specifically with β-amyloid protein. The epitope recognized by the antibody resides within amino acids 1-12 of the β-amyloid protein. It specifically stains amyloid plaques within the cortex and amyloid deposits in blood vessels using formic acid-treated, formalin-fixed, paraffin-embedded, and Methacarn-fixed sections of human Alzheimer′s disease (AD) brain tissue.

Immunogen

Synthetic β-amyloid peptide, conjugated to KLH.

Application

Monoclonal Anti-β-Amyloid antibody has been used in immunoprecipitation analysis, microarray-based immunoassay and microarray.
Mouse monoclonal anti-ABETA was used to treat old WT PDAPP mice with amyloid accumulation and learning deficits in an attempt to improve learning and decrease accumulation, however no response was observed.
The antibody is useful in immunohistochemistry, immunoblotting, ELISA, and competitive ELISA. Also, this antibody has been used to neutralize Aβ assemblies in brains of transgenic mice expressing a mutant form of amyloid precursor protein, and for in vivo deep tissue imaging using near-IR optical spectrum.

Biochem/physiol Actions

β-amyloid gene acts as the substrate of insulin-degrading enzyme (IDE). It plays a major role in the pathogenesis of Alzheimer′s disease (AD) and type 2 diabetes mellitus (DM2).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antibodies targeted to the brain with image-guided focused ultrasound reduces amyloid-β plaque load in the TgCRND8 mouse model of Alzheimer's disease.
Jord?o JF, et al.
PLoS ONE, 5(5), e10549-e10549 (2010)
Alexandra Petiet et al.
Methods in molecular biology (Clifton, N.J.), 771, 293-308 (2011-08-30)
Alzheimer's disease (AD) is an important social and economic issue for our societies. The development of therapeutics against this severe dementia requires assessing the effects of new drugs in animal models thanks to dedicated biomarkers. According to the amyloid cascade
Association of Antineuronal Antibody Levels with Cognitive Impairment in Older Cuban Adults.
Leonardo Cristiá-Lara et al.
MEDICC review, 19(2-3), 32-32 (2017-07-01)
Insulin-degrading enzyme regulates the levels of insulin, amyloid β-protein, and the β-amyloid precursor protein intracellular domain in vivo.
Farris W, et al.
Proceedings of the National Academy of Sciences of the USA, 100(7), 4162-4167 (2003)
Charlotte Gary et al.
Acta neuropathologica communications, 7(1), 126-126 (2019-09-05)
Alzheimer's disease is characterized by cognitive alterations, cerebral atrophy and neuropathological lesions including neuronal loss, accumulation of misfolded and aggregated β-amyloid peptides (Aβ) and tau proteins. Iatrogenic induction of Aβ is suspected in patients exposed to pituitary-derived hormones, dural grafts

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service