Skip to Content
Merck
All Photos(1)

Key Documents

A3650

Sigma-Aldrich

Amikacin hydrate

aminoglycoside antibiotic

Synonym(s):

N1-[(S)-4-Amino-2-hydroxybutyryl]kanamycin A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H43N5O13 · xH2O
CAS Number:
Molecular Weight:
585.60 (anhydrous basis)
Beilstein:
1445422
EC Number:
MDL number:
UNSPSC Code:
51281632
PubChem Substance ID:
NACRES:
NA.85

biological source

synthetic

form

powder

color

white to almost white

antibiotic activity spectrum

Gram-negative bacteria
mycobacteria

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

O.NCC[C@H](O)C(=O)N[C@@H]1C[C@H](N)[C@@H](O[C@H]2O[C@H](CN)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O[C@H]3O[C@H](CO)[C@@H](O)[C@H](N)[C@H]3O

InChI

1S/C22H43N5O13.H2O/c23-2-1-8(29)20(36)27-7-3-6(25)18(39-22-16(34)15(33)13(31)9(4-24)37-22)17(35)19(7)40-21-14(32)11(26)12(30)10(5-28)38-21;/h6-19,21-22,28-35H,1-5,23-26H2,(H,27,36);1H2/t6-,7+,8-,9+,10+,11-,12+,13+,14+,15-,16+,17-,18+,19-,21+,22+;/m0./s1

InChI key

DTSOZYYWEZJFSS-XTHCGPPUSA-N

General description

Chemical structure: aminoglycoside

Application

Amikacin hydrate is used in antimicrobial susceptibility studies of organisms such as Mycobacterium tuberculosis and Ehrlichia phagocytophila.

Biochem/physiol Actions

Amikacin inhibits bacterial protein synthesis by binding to the 30S ribosome subunit and inducing mRNA misreading. It interferes with the translocation of tRNA from the A-site to the P-site.

Packaging

5G

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M P Goren et al.
The Pediatric infectious disease journal, 8(5), 278-282 (1989-05-01)
A three-drug antibiotic regimen including vancomycin and amikacin has been recommended as effective treatment in clinical settings in which Gram-positive bacteremias are a serious problem. To determine if vancomycin potentiates the tubular proteinuria associated with amikacin therapy, we studied febrile
G J Alangaden et al.
Antimicrobial agents and chemotherapy, 42(5), 1295-1297 (1998-05-21)
An A1400G mutation of the rrs gene was identified in Mycobacterium tuberculosis (MTB) strain ATCC 35827 and in 13 MTB clinical isolates resistant to amikacin-kanamycin (MICs, >128 microg/ml). High-level cross-resistance may result from such a mutation since MTB has a
H W Horowitz et al.
Antimicrobial agents and chemotherapy, 45(3), 786-788 (2001-02-22)
Human granulocytic ehrlichiosis is a recently described disease caused by an obligate intracellular gram-negative organism recently named Ehrlichia phagocytophila. To expand our knowledge of the susceptibility of E. phagocytophila, we tested six New York State isolates for susceptibility to 12
Meenu Sharma et al.
Journal of clinical microbiology, 49(12), 4112-4116 (2011-10-15)
The purpose of this study was to establish a standardized protocol for second-line antimicrobial susceptibility testing of Mycobacterium tuberculosis using the Bactec MGIT 960 system in Canadian laboratories. Four Canadian public health laboratories compared the susceptibility testing results of 9
Hessel Van der Weide et al.
Antibiotics (Basel, Switzerland), 9(3) (2020-03-07)
Background: Recent scientific reports on the use of high dose tigecycline monotherapy as a "drug of last resort" warrant further research into the use of this regimen for the treatment of severe multidrug-resistant, Gram-negative bacterial infections. In the current study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service