Skip to Content
Merck
All Photos(1)

Key Documents

A3563

Sigma-Aldrich

Anti-Mouse IgG (whole molecule) F(ab′)2 fragment–Alkaline Phosphatase antibody produced in sheep

affinity isolated antibody, buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46
Pricing and availability is not currently available.

biological source

sheep

Quality Level

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

mouse

technique(s)

direct ELISA: 1:30,000
dot blot: 1:30,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50
western blot: 1:30,000

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Immunoglobulin G (IgG) is a glycoprotein antibody that regulates immune responses such as phagocytosis and is also involved in the development of autoimmune diseases . Mouse IgGs have four distinct isotypes, namely, IgG1, IgG2a, IgG2b, and IgG3. IgG1 regulates complement fixation in mice
Sheep anti-mouse IgG (whole molecule) F(ab′)2 fragment is specific for the F(ab′)2 of mouse IgG. The antibody preparation consists only of the F(ab′)2 fragment of sheep IgG as determined by SDS-PAGE. No contamination with sheep IgG whole molecule is observed.

Immunogen

Mouse IgG

Application

Anti-Mouse IgG (whole molecule) F(ab′)2 fragment-Alkaline Phosphatase antibody is suitable for use in direct ELISA (1:30,000), western blot (1:30,000), and immunohistochemistry (at 1:50 dilution in formalin-fixed, paraffin-embedded sections). The antibody may also be used for dot blot and immunoblot assays using a working dilution of 1:30,000.
ELISAs using alkaline phosphatase conjugated sheep anti-mouse IgG whole molecule antibody were performed to measure the antibody response in the sera of mice immunixed to T. crassiceps. 1:1000 at 37 degrees for one hour and developed using p-nitrophenyl (Sigma).

Physical form

Solution in 0.05 M Tris, pH 8.0, containing 1% bovine serum albumin, 1 mM MgCl2, 50% glycerol, and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rajeev K Tyagi et al.
Scientific reports, 7, 41083-41083 (2017-02-16)
Targeting of myeloid-dendritic cell receptor DC-SIGN by numerous chronic infectious agents, including Porphyromonas gingivalis, is shown to drive-differentiation of monocytes into dysfunctional mDCs. These mDCs exhibit alterations of their fine-tuned homeostatic function and contribute to dysregulated immune-responses. Here, we utilize
Raymond Schuch et al.
PloS one, 8(4), e60754-e60754 (2013-04-18)
We identified an essential cell wall biosynthetic enzyme in Bacillus anthracis and an inhibitor thereof to which the organism did not spontaneously evolve measurable resistance. This work is based on the exquisite binding specificity of bacteriophage-encoded cell wall-hydrolytic lysins, which
Pachiappan Arjunan et al.
Scientific reports, 8(1), 16607-16607 (2018-11-11)
Chronic periodontitis (CP) is a microbial dysbiotic disease linked to increased risk of oral squamous cell carcinomas (OSCCs). To address the underlying mechanisms, mouse and human cell infection models and human biopsy samples were employed. We show that the 'keystone'
Heather J Imrie et al.
PloS one, 7(11), e49816-e49816 (2012-11-28)
Haptoglobin related protein (Hpr) is a key component of trypanosome lytic factors (TLF), a subset of high-density lipoproteins (HDL) that form the first line of human defence against African trypanosomes. Hpr, like haptoglobin (Hp) can bind to hemoglobin (Hb) and
Ittai Fattal et al.
Immunology, 143(3), 374-380 (2014-05-14)
Pemphigus vulgaris (PV) is an autoimmune skin disease, which has been characterized by IgG autoantibodies to desmoglein 3. Here we studied the antibody signatures of PV patients compared with healthy subjects and with patients with two other autoimmune diseases with

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service