Skip to Content
Merck
All Photos(1)

Key Documents

A3506

Sigma-Aldrich

L-(−)-Arabitol

≥98% (GC)

Synonym(s):

L-Arabinitol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12O5
CAS Number:
Molecular Weight:
152.15
Beilstein:
1720521
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98% (GC)

form

powder

color

white

mp

101-104 °C (lit.)

solubility

water: 50 mg/mL, clear, colorless

SMILES string

OC[C@H](O)C(O)[C@@H](O)CO

InChI

1S/C5H12O5/c6-1-3(8)5(10)4(9)2-7/h3-10H,1-2H2/t3-,4-/m0/s1

InChI key

HEBKCHPVOIAQTA-IMJSIDKUSA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-Arabitol, a rare sugar alcohol, is being studied as a food additive that reduces fat deposits in the intestines. L-Arabitol is used as an inducer of xylanase expression in Hypocrea jecorina (Trichoderma reesei). L-Arabitol is used to identify, differentiate and characterize L-arabitol dehydrogenase(s).

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Onkenhout et al.
Molecular genetics and metabolism, 77(1-2), 80-85 (2002-10-03)
A female patient, the first child of healthy non-consanguineous parents, presented at the age of 16 months with delayed motor development and facial dysmorphism. In addition she displayed a palatoschizis and multiple skeletal abnormalities as hypoplastic scapulae, hypoplastic os ilea
Gregory D Tredwell et al.
PloS one, 6(1), e16286-e16286 (2011-02-02)
Metabolic profiling is increasingly being used to investigate a diverse range of biological questions. Due to the rapid turnover of intracellular metabolites it is important to have reliable, reproducible techniques for sampling and sample treatment. Through the use of non-targeted
Dominik Mojzita et al.
The Journal of biological chemistry, 285(31), 23622-23628 (2010-06-01)
The first enzyme in the pathway for l-arabinose catabolism in eukaryotic microorganisms is a reductase, reducing l-arabinose to l-arabitol. The enzymes catalyzing this reduction are in general nonspecific and would also reduce d-xylose to xylitol, the first step in eukaryotic
Barney A Geddes et al.
Microbiology (Reading, England), 158(Pt 8), 2180-2191 (2012-05-23)
The Sinorhizobium meliloti locus necessary for the utilization of erythritol as a sole carbon source, contains 17 genes, including genes that encode an ABC transporter necessary for the transport of erythritol, as well as the genes encoding EryA, EryB, EryC
Astrid R Mach-Aigner et al.
Applied and environmental microbiology, 77(17), 5988-5994 (2011-07-12)
The saprophytic fungus Hypocrea jecorina (anamorph, Trichoderma reesei) is an important native producer of hydrolytic enzymes, including xylanases. Regarding principles of sustainability, cheap and renewable raw materials, such as d-xylose (the backbone monomer of xylan), have been receiving increasing attention

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service