Skip to Content
Merck
All Photos(1)

Key Documents

A2562

Sigma-Aldrich

Angiotensinogen from human plasma

≥80% (SDS-PAGE)

Synonym(s):

Hypertensinogen, Renin substrate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.26

Product Name

Angiotensinogen from human plasma, ≥80% (SDS-PAGE), lyophilized powder

Quality Level

Assay

≥80% (SDS-PAGE)

form

lyophilized powder

color

white

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... AGT(183)

Biochem/physiol Actions

Angiotensinogen (renin substrate), a component of the rennin-angiotensin system, is a precursor protein to a family of blood pressure and hormone release regulating peptides known as the angiotensins.

Packaging

Package size based on protein content.

Physical form

Lyophilized powder containing sodium phosphate buffer, pH 7.

Analysis Note

Protein determined by A1%/280

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Alin Ciobica et al.
Acta neurologica Belgica, 109(3), 171-180 (2009-11-12)
Biochemical, physiological and functional studies suggest that the brain renin-angiotensin system (RAS) is regulated independently of the peripheral RAS. Also, all enzymes and peptides necessary for the biosynthesis of various angiotensins have been recognized within the central nervous system. There
Weijian Shao et al.
American journal of physiology. Renal physiology, 304(5), F505-F514 (2013-01-11)
In angiotensin II (ANG II) infusion hypertension, there is an augmentation of intratubular angiotensinogen (AGT) and ANG II leading to increased urinary AGT and ANG II excretion rates associated with tissue injury. However, the changes in urinary AGT and ANG
Jie Yang et al.
Journal of molecular endocrinology, 50(2), R53-R66 (2013-02-02)
The compensatory alterations in the rennin-angiotensin-aldosterone system (RAAS) contribute to the salt-water balance and sufficient placental perfusion for the subsequent well-being of the mother and fetus during normal pregnancy and is characterized by an increase in almost all the components
Angiotensinogen gene polymorphism and HDL2 are linked to coronary artery calcification in individuals with family history of early coronary disease.
Andrei C Sposito et al.
Atherosclerosis, 226(2), 339-340 (2013-01-01)
Lodi C W Roksnoer et al.
Current hypertension reports, 15(2), 81-88 (2013-01-09)
Recent interest focuses on urinary renin and angiotensinogen as markers of renal renin-angiotensin system activity. Before concluding that these components are independent markers, we need to exclude that their presence in urine, like that of albumin (a protein of comparable

Articles

Lipid Induced Insulin Resistance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service