Skip to Content
Merck
All Photos(2)

Documents

A25120

Sigma-Aldrich

DEAE–Sephadex® A-25 chloride form

Synonym(s):

DEAE Sephadex®, Sephadex A 25,2-(diethylamino)ethylether, Diethylaminoethyl Sephadex®

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
47101511
NACRES:
NA.56

form

powder

Quality Level

technique(s)

activity assay: suitable

matrix

cross-linked dextran

matrix active group

functional group

bead size

40-125 μm (dry)

pore size

~30,000 Da exclusion limit

operating pH

2-9

capacity

3-4 meq/g ion exchange capacity

compatibility

mode of use mixed weak and strong anion exchange chromatography

General description

The Sephadex® ion exchangers contain a cross-linked dextran matrix with functional groups. These groups are linked by stable ether linkages to the glucose units in the matrix. DEAE Sephadex can be used in ion exchange chromatography for purifying and isolating proteins.

A25120-100G′s updated product number is GE17-0170-01
A25120-500G′s updated product number is GE17-0180-02

Application

DEAE-Sephadex® A-25 has been used:
  • to form DEAE-Sephadex A-25 (formate form) for glucosinolates (GLS) extraction using horseradish plants
  • in the estimation of GLS content in Chinese kale sprouts
  • production and purification of endozepine from astrocytes
  • to identify and label an insulin-activated nitric oxide synthase inhibitor (protein) in acute myocardial infarction (AMI) patients

Features and Benefits

  • Weak anion exchanger
  • Suitable for batch techniques
  • High binding capacities
  • Non-reactive to commonly used aqueous buffers additives such as 8 M urea and 6 M guanidine hydrochloride

Legal Information

DEAE Sephadex is a registered trademark of Cytiva
Sephadex is a registered trademark of Cytiva

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Human Lymphocyte Agglutinins in <I>Tursiops Truncatus</I> (Bottlenose Dolphin).
Hohn, D., et al.
Immunological Investigations, 12(5), 481-490 (1983)
T M Parkinson
Journal of lipid research, 8(1), 24-29 (1967-01-01)
Various cellulose and dextran anion exchangers bind bile salts in vitro under conditions of pH and ionic strength resembling those in the lumen of the small intestine. Of these substances, diethylaminoethyl (DEAE) cellulose, guanidoethyl cellulose, and DEAE Sephadex reduced hypercholesterolemia
A C Newton et al.
Analytical biochemistry, 156(1), 56-60 (1986-07-01)
The greatly enhanced fluorescence of Tb3+ when complexed with dipicolinic acid affords a simple and highly sensitive method for monitoring continuous anion flux through the erythrocyte anion transporter, band 3. Dipicolinic acid (pyridine-2,6-dicarboxylic acid) is entrapped in human erythrocytes and
M J Hollingsworth et al.
Molecular and cellular biology, 6(4), 1058-1064 (1986-04-01)
A requisite step in the biosynthesis of tRNA is the removal of 5' leader sequences from tRNA precursors. We have detected an RNase P activity in yeast mitochondrial extracts that can carry out this reaction on a homologous precursor tRNA.
Scott N Boyle et al.
Biochemistry, 46(41), 11614-11620 (2007-09-26)
Abl family kinases have been implicated in the regulation of cell morphogenesis and migration, but the molecular mechanisms through which they operate are not fully elucidated. We applied the bump-hole technique, pioneered by Shokat and colleagues, to identify direct substrates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service