Skip to Content
Merck
All Photos(1)

Key Documents

A1727

Sigma-Aldrich

Alkaline Hypochlorite solution

Synonym(s):

Hypochlorite, Sodium Hypochlorite

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352106
NACRES:
NA.25

form

liquid

Quality Level

storage temp.

2-8°C

Application

Alkaline Hypochlorite solution is used as an oxidizing and bleaching agent with activity as a discolorant.

Components

0.2% sodium hypochlorite in alkali solution

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Haenni et al.
International endodontic journal, 36(2), 100-105 (2003-03-27)
Chemical and antimicrobial effects of paste-like suspensions of calcium hydroxide (Ca(OH)2) powder with chlorhexidine (CHx), sodium hypochlorite (NaOCl) or iodine potassium iodide (IPI) solutions were tested and compared to conventional Ca(OH)2/saline paste. Equivalent mixtures of Ca(OH)2 with irrigating solutions were
D M Taylor et al.
Archives of virology, 139(3-4), 313-326 (1994-01-01)
Macerates of bovine brain infected with bovine spongiform encephalopathy (BSE) agent, and rodent brain infected with the 263K or ME7 strains of scrapie agent, were subjected to porous-load autoclaving at temperatures between 134 and 138 degrees C for < or
Karin Lemmer et al.
The Journal of general virology, 85(Pt 12), 3805-3816 (2004-11-24)
Effective reprocessing of surgical instruments ensuring elimination of inadvertent contamination with infectious agents causing transmissible spongiform encephalopathies (TSEs) is essential for the prevention of iatrogenic transmission of Creutzfeldt-Jakob disease (CJD) or its new variant (vCJD) from asymptomatic carriers. In a
Alberto Amaretti et al.
Frontiers in microbiology, 10, 2614-2614 (2019-12-06)
Unabsorbed proteins reach the colon and are fermented by the microbiota, yielding a variety of harmful metabolites. In the present study, a 16S rRNA gene survey identified the bacterial taxa flourishing in 11 batch fermentations with proteins and peptones as
Suzanne Z Andersen et al.
Nature, 570(7762), 504-508 (2019-05-23)
The electrochemical synthesis of ammonia from nitrogen under mild conditions using renewable electricity is an attractive alternative1-4 to the energy-intensive Haber-Bosch process, which dominates industrial ammonia production. However, there are considerable scientific and technical challenges5,6 facing the electrochemical alternative, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service