Skip to Content
Merck
All Photos(2)

Key Documents

A1682

Sigma-Aldrich

Anti-Mouse IgG (Fab specific)–Alkaline Phosphatase antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Goat Anti-Mouse IgG (Fab specific)–AP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse

should not react with

human, rat

technique(s)

direct ELISA: 1:40,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:10
western blot: 1:200,000-1:400.000 using assay detecting β-Actin in total cell extract of HeLa cells (5-10 μg per well)

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Immunoglobulin G (IgG) is a glycoprotein antibody that regulates immune responses such as phagocytosis and is also involved in the development of autoimmune diseases . Mouse IgGs have four distinct isotypes, namely, IgG1, IgG2a, IgG2b, and IgG3. IgG1 regulates complement fixation in mice . Anti-Mouse IgG (Fab specific)-Alkaline Phosphatase antibody detects mouse IgG (whole molecule). When tested by ELISA, the product reacts with the Fab fragment of mouse IgG but does not recognize the Fc fragment of mouse IgG, human IgG, IgA, IgM or rat IgG.

Immunogen

Purified mouse IgG

Application

Alkaline phosphatase-conjugated goat anti-mouse IgG (Fab specific) was used as the secondary antibody in ELISA assays to measure the reacitivity of mABs to thyrogobulin peptides. Absorbance was measured at 405 nm.
Anti-Mouse IgG (Fab specific)-Alkaline Phosphatase antibody is suitable for use in western blot at 1:1000 dilution using human airway epithelial cells or using whole-cell extracts of rat tubal tissues . The antibody can also be used for immunohistochemistry (1:10 using formalin-fixed, paraffin-embedded sections).
Goat anti-mouse Fab specific alkaline phosphatase antibody was used to analyze serum Ig titers by His Sorb ELISA at 1:10000 at room temperature for two hours. Antibodies were diluted in PBS plus 0.2% BSA to reduce background.

Other Notes

Antibody adsorbed with human IgG and rat serum proteins.

Physical form

Solution in 0.05 M Tris, pH 8.0, containing 1% bovine serum albumin, 1 mM MgCl2 with 50% glycerol as stabilizer and 15 mM sodium azide

Preparation Note

Adsorbed to reduce background staining with human or rat samples.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shweta R Urva et al.
Journal of pharmaceutical sciences, 99(3), 1582-1600 (2009-09-24)
Antibodies directed against tumor associated antigens are being increasingly used for detection and treatment of cancers; however, there is an incomplete understanding of the physiological determinants of antibody pharmacokinetics and tumor distribution. The purpose of this study is to (a)
Ruijin Shao et al.
Biology of reproduction, 80(6), 1262-1271 (2009-02-13)
Clomiphene citrate (CC) therapy for disorders of anovulatory infertility has been linked to an increased frequency of tubal ectopic pregnancy. Although CC enhances apoptotic processes in the ovaries, villi, and decidual tissues, its effect on apoptosis in the fallopian tube
Ruijin Shao et al.
Biology of reproduction, 79(4), 748-757 (2008-07-04)
The anterior pituitary-derived hormone prolactin (PRL) signals through the PRL receptor (PRLR) and is important for female reproductive function in mammals. In contrast to the extensive studies of PRLR expression and regulation in human and mouse ovary and uterus, the
Patrick L Sinn et al.
Journal of virology, 77(10), 5902-5910 (2003-04-30)
The practical application of gene therapy as a treatment for cystic fibrosis is limited by poor gene transfer efficiency with vectors applied to the apical surface of airway epithelia. Recently, folate receptor alpha (FR alpha), a glycosylphosphatidylinositol-linked surface protein, was
Mohamad Zaidan et al.
JCI insight, 5(9) (2020-05-08)
The loss of functional nephrons after kidney injury triggers the compensatory growth of the remaining ones to allow functional adaptation. However, in some cases, these compensatory events activate signaling pathways that lead to pathological alterations and chronic kidney disease. Little

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service