Skip to Content
Merck
All Photos(2)

Key Documents

A1127

Sigma-Aldrich

Adenosine 5′-monophosphomorpholidate 4-morpholine-N,N′-dicyclohexylcarboxamidine salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H21N6O7P · C17H31N3O
CAS Number:
Molecular Weight:
709.77
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

natural (organic)

Assay

≥93% (HPLC)

form

powder

solubility

water: 50 mg/mL, clear, colorless to light yellow

storage temp.

−20°C

SMILES string

C1CCC(CC1)N\C(=N\C2CCCCC2)N3CCOCC3.Nc4ncnc5n(cnc45)[C@@H]6O[C@H](COP(O)(=O)N7CCOCC7)[C@@H](O)[C@H]6O

InChI

1S/C17H31N3O.C14H21N6O7P/c1-3-7-15(8-4-1)18-17(20-11-13-21-14-12-20)19-16-9-5-2-6-10-16;15-12-9-13(17-6-16-12)20(7-18-9)14-11(22)10(21)8(27-14)5-26-28(23,24)19-1-3-25-4-2-19/h15-16H,1-14H2,(H,18,19);6-8,10-11,14,21-22H,1-5H2,(H,23,24)(H2,15,16,17)/t;8-,10-,11-,14-/m.1/s1

InChI key

VYSFOCCDQZWOQI-HZGXGGOMSA-N

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service